Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • RAC1/CDC42 Antibodies

          Invitrogen

          Phospho-RAC1/CDC42 (Ser71) Polyclonal Antibody

          View all (6) RAC1/CDC42 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-RAC1/CDC42 (Ser71) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          Phospho-RAC1/CDC42 (Ser71) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-RAC1/CDC42 (Ser71) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-RAC1/CDC42 (Ser71) Antibody (PA5-85105) in ICC/IF

          Phospho-RAC1/CDC42 (Ser71) Polyclonal Antibody detects Rac1 protein at cytoskeleton by immunofluorescent analysis. Sample: A431 cells were fixed in 4% paraformaldehyde at RT for 15 min. Green: Rac1 stained by Phospho-RAC1/CDC42 (Ser71) Polyclonal Antibody (Product # PA5-85105) diluted at 1:500. Blue: Fluoroshield with DAPI . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-RAC1/CDC42 (Ser71) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-RAC1/CDC42 (Ser71) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RAC1/CDC42 (Ser71) Antibody in Western Blot (WB)
          Phospho-RAC1/CDC42 (Ser71) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-85105

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human Rac1. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.79 mg/mL

          Amount

          79 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2792253

          Product Specific Information

          Keep as concentrated solution.

          Predicted reactivity: Mouse (97%), Rat (90%), Drosophila (82%), Xenopus laevis (90%), Dog (90%), Chicken (90%), Sheep (90%), Chimpanzee (90%), Bovine (90%).

          Positive Control: 293T, A431, HeLa, HepG2, Molt-4, Raji, mouse liver.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; cell migration-inducing gene 5 protein; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; GTP-binding protein G25K; p21-Rac1; Rac1ins; ras-like protein; ras-like protein TC25; ras-related C3 botulinum toxin substrate; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; small GTP binding protein; small GTP binding protein CDC42; small GTP binding protein; GTPase; small GTP binding protein; Rac1; Rho-related protein; unnamed protein product

          View more View less

          Gene Aliases: CDC42Hs; G25K; MIG5; MRD48; p21-Rac1; Rac-1; TC-25; TKS

          View more View less

          UniProt ID: (Human) P60953, (Rat) Q8CFN2, (Human) P63000, (Rat) Q6RUV5

          View more View less

          Entrez Gene ID: (Human) 998, (Rat) 64465, (Human) 5879, (Rat) 363875

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding protein kinase binding GTP-dependent protein binding thioesterase binding GBD domain binding apolipoprotein A-I receptor binding identical protein binding ubiquitin protein ligase activity mitogen-activated protein kinase kinase kinase binding enzyme binding macromolecular complex binding Rho GDP-dissociation inhibitor binding small GTPase binding histone deacetylase binding ATPase binding
          Process(es)
          heart process cardiac neural crest cell migration involved in outflow tract morphogenesis endocytosis phagocytosis, engulfment actin filament organization Golgi organization establishment or maintenance of cell polarity signal transduction integrin-mediated signaling pathway small GTPase mediated signal transduction anatomical structure morphogenesis regulation of lamellipodium assembly positive regulation of lamellipodium assembly substantia nigra development establishment of cell polarity actin cytoskeleton organization macrophage differentiation positive regulation of cell growth positive regulation of cell migration positive regulation of pseudopodium assembly negative regulation of protein complex assembly positive regulation of cytokinesis regulation of actin cytoskeleton organization cell junction assembly embryonic heart tube development neuropilin signaling pathway modulation by host of viral process establishment of epithelial cell apical/basal polarity positive regulation of pinocytosis positive regulation of cellular component organization regulation of filopodium assembly positive regulation of filopodium assembly regulation of stress fiber assembly positive regulation of stress fiber assembly establishment of Golgi localization regulation of attachment of spindle microtubules to kinetochore Wnt signaling pathway, planar cell polarity pathway positive regulation of epithelial cell proliferation involved in lung morphogenesis dendritic spine morphogenesis organelle transport along microtubule endothelin receptor signaling pathway regulation of postsynapse organization positive regulation of substrate adhesion-dependent cell spreading sprouting angiogenesis cardiac conduction system development protein phosphorylation regulation of mitotic nuclear division nuclear migration protein localization response to glucose regulation of exocytosis adherens junction organization establishment or maintenance of apical/basal cell polarity dendritic cell migration positive regulation of catalytic activity positive regulation of MAPK cascade positive regulation of neuron apoptotic process positive regulation of DNA replication positive regulation of JNK cascade filopodium assembly neuron fate determination nucleus localization positive regulation of synapse structural plasticity positive regulation of protein kinase B signaling heart contraction submandibular salivary gland formation cellular response to interferon-gamma actin filament branching positive regulation of intracellular protein transport regulation of modification of postsynaptic structure neuron migration positive regulation of protein phosphorylation sphingosine-1-phosphate signaling pathway inflammatory response cell adhesion cell-matrix adhesion enzyme linked receptor protein signaling pathway motor neuron axon guidance regulation of cell shape response to wounding regulation of hydrogen peroxide metabolic process positive regulation of endothelial cell migration regulation of fibroblast migration negative regulation of fibroblast migration regulation of cell-substrate adhesion cell migration Rac protein signal transduction cell projection assembly lamellipodium assembly actin filament polymerization regulation of cell migration cortical cytoskeleton organization forebrain development ruffle organization response to lipopolysaccharide negative regulation of interleukin-23 production substrate adhesion-dependent cell spreading positive regulation of Rho protein signal transduction intracellular signal transduction non-canonical Wnt signaling pathway positive regulation of insulin secretion involved in cellular response to glucose stimulus superoxide anion generation engulfment of apoptotic cell regulation of nitric oxide biosynthetic process respiratory burst regulation of JAK-STAT cascade hepatocyte growth factor receptor signaling pathway negative regulation of receptor-mediated endocytosis neuron projection morphogenesis cell motility localization within membrane positive regulation of focal adhesion assembly regulation of respiratory burst cell chemotaxis regulation of cell adhesion involved in heart morphogenesis semaphorin-plexin signaling pathway positive regulation of neutrophil chemotaxis ruffle assembly regulation of postsynapse assembly positive regulation of ruffle assembly regulation of synaptic vesicle endocytosis regulation of neutrophil migration positive regulation of bicellular tight junction assembly midbrain dopaminergic neuron differentiation positive regulation of ovarian follicle development auditory receptor cell morphogenesis mast cell chemotaxis epithelial cell morphogenesis chemotaxis hyperosmotic response cytoskeleton organization G-protein coupled receptor signaling pathway axon guidance regulation of neuron maturation dendrite development cerebral cortex radially oriented cell migration embryonic olfactory bulb interneuron precursor migration cerebral cortex GABAergic interneuron development regulation of cell morphogenesis positive regulation of actin filament polymerization angiotensin-activated signaling pathway erythrocyte enucleation cell-cell junction organization bone resorption regulation of neuronal synaptic plasticity anatomical structure arrangement dendrite morphogenesis multicellular organismal homeostasis homeostasis of number of cells within a tissue synaptic transmission, GABAergic positive regulation of dendritic spine development regulation of ERK5 cascade cellular response to mechanical stimulus dopaminergic neuron differentiation protein localization to plasma membrane cochlea morphogenesis postsynaptic actin cytoskeleton organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-xtlnb:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0