Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • RUNX1 Antibodies

          Invitrogen

          Phospho-RUNX1 (Ser249) Polyclonal Antibody

          View all (41) RUNX1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-RUNX1 (Ser249) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (40)
          Phospho-RUNX1 (Ser249) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-RUNX1 (Ser249) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 40

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-RUNX1 (Ser249) Antibody (PA5-104570) in ICC/IF

          Immunocytochemistry analysis of Phospho-RUNX1 (Ser249) in Heat-shock treated Hela cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with Phospho-RUNX1 (Ser249) polyclonal antibody (Product # PA5-104570) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-RUNX1 (Ser249) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RUNX1 (Ser249) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-RUNX1 (Ser249) Polyclonal Antibody

          Product Details

          PA5-104570

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human RUNX1(Accession Q01196), corresponding to amino acid residues around phosphorylated Ser249.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816045

          Product Specific Information

          Antibody detects endogenous levels of RUNX1 / AML1 only when phosphorylated at Ser249.

          Target Information

          RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: acute myeloid leukemia 1 protein; AML-1; AML1; aml1 oncogene; AML1-ETO fusion; AML1-ETO fusion protein; AML1-EVI-1 fusion protein; CBF-alpha 2; CBFA2; core binding factor alpha 2; core-binding factor runt domain alpha subunit 2 (acute myeloid leukemia 1 oncogene); core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; mutant RUNX1; oncogene AML-1; OTTHUMP00000108697; OTTHUMP00000108700; OTTHUMP00000108702; PEA2-alpha B; PEBP2-alpha B; polyomavirus enhancer-binding protein 2 alpha B subunit; RUN1; runt domain, alpha subunit 2; runt related transcription factor 1; RUNX1 with skipped exon 5; similar to Homo sapiens AML1c; acute myeloid leukemia 1; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit; unnamed protein product

          View more View less

          Gene Aliases: AML1; AML1-EVI-1; AMLCR1; CBF-alpha-2; CBF2alpha; CBFA2; EVI-1; PEA2-alpha; PEBP2-alpha; Pebp2a2; PEBP2aB; PEBP2alpha; Pebpa2b

          View more View less

          UniProt ID: (Human) Q01196, (Rat) Q63046

          View more View less

          Entrez Gene ID: (Human) 861, (Mouse) 12394, (Rat) 50662

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription corepressor binding transcription coactivator binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding calcium ion binding protein binding ATP binding protein homodimerization activity protein heterodimerization activity DNA-binding transcription factor binding sequence-specific double-stranded DNA binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ossification chondrocyte differentiation myeloid leukocyte differentiation regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter regulation of plasminogen activation hemopoiesis myeloid cell differentiation neuron differentiation negative regulation of granulocyte differentiation positive regulation of granulocyte differentiation positive regulation of interleukin-2 production positive regulation of collagen biosynthetic process negative regulation of CD4-positive, alpha-beta T cell differentiation positive regulation of CD8-positive, alpha-beta T cell differentiation regulation of cell differentiation positive regulation of angiogenesis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter peripheral nervous system neuron development positive regulation of developmental process positive regulation of multicellular organismal process regulation of cardiac muscle cell proliferation cardiac muscle tissue regeneration hematopoietic stem cell proliferation positive regulation of extracellular matrix organization regulation of connective tissue replacement regulation of multicellular organismal development skeletal system development in utero embryonic development liver development myeloid progenitor cell differentiation regulation of T cell anergy central nervous system development negative regulation of cell proliferation regulation of signal transduction gene expression hair follicle morphogenesis response to retinoic acid positive regulation of interferon-gamma production embryonic hemopoiesis negative regulation of transcription, DNA-templated behavioral response to pain cell development neuron fate commitment neuron development definitive hemopoiesis regulation of hair follicle cell proliferation cellular response to transforming growth factor beta stimulus positive regulation of cell maturation positive regulation of progesterone secretion response to denervation involved in regulation of muscle adaptation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • 隱私權聲明
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch