Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PKD2 Antibodies

          Bethyl Laboratories

          Polycystin-2 Polyclonal Antibody

          View all (19) PKD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Polycystin-2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Polycystin-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Polycystin-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Polycystin-2 Antibody (A302-470A-T) in WB

          Detection of mouse Polycystin-2 by western blot. Samples: Whole cell lysate (50 µg) from TCMK-1 and NIH 3T3 cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-Polycystin-2 antibody (Product # A302-470A) (lot A302-470A-3) used for WB at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 75 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Polycystin-2 Antibody in Western Blot (WB)
          Polycystin-2 Antibody in Western Blot (WB)
          Polycystin-2 Antibody in Immunoprecipitation (IP)
          Polycystin-2 Polyclonal Antibody

          Product Details

          A302-470A-T

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          -

          Immunoprecipitation (IP)

          2-5 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 918 and 968 of Human polycystin-2.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          10 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris citrate/phosphate, pH 7-8

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Target Information

          This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal dominant polycystic kidney disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A MGC138466; autosomal dominant polycystic kidney disease type II; Autosomal dominant polycystic kidney disease type II protein; MGC138468; PC2; PKD2; polycystic kidney disease 2 (autosomal dominant); Polycystic kidney disease 2 protein; Polycystic kidney disease 2 protein homolog; Polycystin 2; Polycystin-2; Polycystwin; R48321; Transient receptor potential cation channel subfamily P member 2; TRPP1; unnamed protein product

          View more View less

          Gene Aliases: APKD2; C030034P18Rik; Pc-2; PC2; PKD2; PKD4; TRPP1; TRPP2

          View more View less

          UniProt ID: (Human) Q13563, (Mouse) O35245

          View more View less

          Entrez Gene ID: (Human) 5311, (Mouse) 18764

          View more View less

          Function(s)
          receptor binding voltage-gated ion channel activity voltage-gated calcium channel activity voltage-gated sodium channel activity voltage-gated potassium channel activity cation channel activity calcium channel activity potassium channel activity sodium channel activity calcium ion binding protein binding cytoskeletal protein binding channel activity outward rectifier potassium channel activity voltage-gated cation channel activity identical protein binding protein homodimerization activity actinin binding HLH domain binding ion channel binding metal ion binding calcium-induced calcium release activity ATPase binding phosphoprotein binding muscle alpha-actinin binding transcription regulator inhibitor activity alpha-actinin binding ion channel
          Process(es)
          branching involved in ureteric bud morphogenesis kidney development liver development embryonic placenta development heart looping detection of nodal flow ion transport potassium ion transport sodium ion transport calcium ion transport cellular calcium ion homeostasis cell surface receptor signaling pathway JAK-STAT cascade determination of left/right symmetry heart development negative regulation of cell proliferation positive regulation of gene expression Wnt signaling pathway calcium-mediated signaling spinal cord development neural tube development ion transmembrane transport cellular response to reactive oxygen species metanephric part of ureteric bud development sodium ion transmembrane transport aorta development regulation of cell proliferation chordate embryonic development cilium organization positive regulation of nitric oxide biosynthetic process positive regulation of transcription from RNA polymerase II promoter embryonic organ development detection of mechanical stimulus release of sequestered calcium ion into cytosol protein tetramerization protein homotetramerization protein heterotetramerization centrosome duplication establishment of localization in cell regulation of cell cycle placenta blood vessel development renal tubule morphogenesis renal artery morphogenesis calcium ion transmembrane transport cellular response to calcium ion cellular response to cAMP cellular response to hydrostatic pressure cellular response to osmotic stress cellular response to fluid shear stress potassium ion transmembrane transport determination of liver left/right asymmetry metanephric mesenchyme development mesonephric tubule development mesonephric duct development metanephric smooth muscle tissue development metanephric cortex development metanephric ascending thin limb development metanephric cortical collecting duct development metanephric distal tubule development metanephric S-shaped body morphogenesis regulation of calcium ion import inorganic cation transmembrane transport positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway negative regulation of G1/S transition of mitotic cell cycle transport cell cycle arrest positive regulation of cytosolic calcium ion concentration regulation of cAMP metabolic process positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle cytoplasmic sequestering of transcription factor transmembrane transport negative regulation of ryanodine-sensitive calcium-release channel activity positive regulation of cell cycle arrest renal system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline