PA1-74090 detects Prothrombin Fragments from human samples.
PA1-74090 has been successfully used in ELISA applications.
The PA1-74090 immunogen is Prothrombin from human plasma.
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.