Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • QRICH1 Antibodies

          Proteintech

          QRICH1 Polyclonal Antibody

          View all (4) QRICH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite QRICH1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (14)
          QRICH1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          QRICH1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          QRICH1 Antibody (30236-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using QRICH1 antibody (30236-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          QRICH1 Antibody in Immunocytochemistry (ICC/IF)
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          QRICH1 Antibody in Western Blot (WB)
          QRICH1 Antibody in Western Blot (WB)
          QRICH1 Polyclonal Antibody

          Product Details

          30236-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:800
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:800
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          QRICH1 fusion protein Ag31997
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.7 mg/mL

          Amount

          105 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MNNSLENTIS FEEYIRVKAR SVPQHRMKEF LDSLASKGPE ALQEFQQTAT TTMVYQQGGN CIYTDSTEVA GSLLELACPV TTSVQPQTQQ EQQIQVQQPQ QVQVQVQVQQ SPQQVSAQLS PQLTVHQPTE QPIQVQVQIQ GQAPQSAAPS IQTPSLQSPS PSQLQAAQIQ VQHVQAAQQI QAAEIPEEHI PHQQIQAQLV AGQSLAGGQQ IQIQTVGALS PPPSQQGSPR EGERRVGTAS VLQPVKKRKV DMPITVSYAI SGQPVATVLA IPQGQQQSYV SLRPDLLTVD SAHLYSATGT

          Target Information

          Glutamine-rich protein 1 contains 1 CARD domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ20259; Glutamine-rich protein 1; MGC131838; Transcriptional regulator QRICH1; unnamed protein product

          View more View less

          Gene Aliases: 2610028H07Rik; AB-DIP; QRICH1; RGD1311095; VERBRAS

          View more View less

          UniProt ID: (Human) Q2TAL8, (Mouse) Q3UA37

          View more View less

          Entrez Gene ID: (Human) 54870, (Rat) 301004, (Mouse) 69232

          View more View less

          Function(s)
          DNA binding protein binding molecular_function zinc finger transcription factor
          Process(es)
          response to unfolded protein endoplasmic reticulum unfolded protein response response to endoplasmic reticulum stress PERK-mediated unfolded protein response positive regulation of apoptotic process positive regulation of transcription, DNA-templated intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress integrated stress response signaling biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline