Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • QRICH1 Antibodies

          Abnova

          QRICH1 Polyclonal Antibody, MaxPab™

          View all (4) QRICH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite QRICH1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          QRICH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          QRICH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          QRICH1 Antibody (H00054870-B01P) in WB

          Western Blot analysis of QRICH1 expression in transfected 293T cell line by QRICH1 MaxPab polyclonal antibody. Lane 1: QRICH1 transfected lysate(85.36 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          QRICH1 Antibody in Western Blot (WB)
          QRICH1 Polyclonal Antibody, MaxPab™

          Product Details

          H00054870-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          QRICH1 (NP_060200.2, 1 a.a. approximately 776 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MNNSLENTIS FEEYIRVKAR SVPQHRMKEF LDSLASKGPE ALQEFQQTAT TTMVYQQGGN CIYTDSTEVA GSLLELACPV TTSVQPQTQQ EQQIQVQQPQ QVQVQVQVQQ SPQQVSAQLS PQLTVHQPTE QPIQVQVQIQ GQAPQSAAPS IQTPSLQSPS PSQLQAAQIQ VQHVQAAQQI QAAEIPEEHI PHQQIQAQLV AGQSLAGGQQ IQIQTVGALS PPPSQQGSPR EGERRVGTAS VLQPVKKRKV DMPITVSYAI SGQPVATVLA IPQGQQQSYV SLRPDLLTVD SAHLYSATGT ITSPTGETWT IPVYSAQPRG DPQQQSITHI AIPQEAYNAV HVSGSPTALA AVKLEDDKEK MVGTTSVVKN SHEEVVQTLA NSLFPAQFMN GNIHIPVAVQ AVAGTYQNTA QTVHIWDPQQ QPQQQTPQEQ TPPPQQQQQQ LQVTCSAQTV QVAEVEPQSQ PQPSPELLLP NSLKPEEGLE VWKNWAQTKN AELEKDAQNR LAPIGRRQLL RFQEDLISSA VAELNYGLCL MTREARNGEG EPYDPDVLYY IFLCIQKYLF ENGRVDDIFS DLYYVRFTEW LHEVLKDVQP RVTPLGYVLP SHVTEEMLWE CKQLGAHSPS TLLTTLMFFN TKYFLLKTVD QHMKLAFSKV LRQTKKNPSN PKDKSTSIRY LKALGIHQTG QKVTDDMYAE QTENPENPLR CPIKLYDFYL FKCPQSVKGR NDTFYLTPEP VVAPNSPIWY SVQPISREQM GQMLTRILVI REIQEAIAVA NASTMH

          Target Information

          Glutamine-rich protein 1 contains 1 CARD domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ20259; Glutamine-rich protein 1; MGC131838; Transcriptional regulator QRICH1; unnamed protein product

          View more View less

          Gene Aliases: AB-DIP; QRICH1; VERBRAS

          View more View less

          UniProt ID: (Human) Q2TAL8

          View more View less

          Entrez Gene ID: (Human) 54870

          View more View less

          Function(s)
          DNA binding protein binding
          Process(es)
          response to unfolded protein endoplasmic reticulum unfolded protein response response to endoplasmic reticulum stress PERK-mediated unfolded protein response positive regulation of apoptotic process positive regulation of transcription, DNA-templated intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress integrated stress response signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline