Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAB27B Antibodies

          Proteintech

          RAB27B Polyclonal Antibody

          50 References
          View all (25) RAB27B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RAB27B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (22)
          RAB27B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB27B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB27B Antibody (13412-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 4% PFA) fixed HeLa cells using 13412-1-AP (RAB27B antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB27B Antibody in Immunocytochemistry (ICC/IF)
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAB27B Antibody in Western Blot (WB)
          RAB27B Antibody in Western Blot (WB)
          RAB27B Antibody in Western Blot (WB)
          RAB27B Antibody in Western Blot (WB)
          RAB27B Antibody in Western Blot (WB)
          RAB27B Polyclonal Antibody

          Product Details

          13412-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 30 publications 30 publications

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 7 publications 7 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAB27B Fusion Protein Ag4064 (1-218 aa encoded by BC027474)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MTDGDYDYL IKLLALGDSG VGKTTFLYRY TDNKFNPKFI TTVGIDFREK RVVYNAQGPN GSSGKAFKVH LQLWDTAGQE RFRSLTTAFF RDAMGFLLMF DLTSQQSFLN VRNWMSQLQA NAYCENPDIV LIGNKADLPD QREVNERQAR ELADKYGIPY FETSAATGQN VEKAVETLLD LIMKRMEQCV EKTQIPDTVN GGNSGNLDGE KPPEKKCIC (1-218 aa encoded by BC027474 )

          Target Information

          RAB27B may be involved in targeting uroplakins to urothelial apical membranes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C25KG; Ras-related protein Rab-27B; unnamed protein product

          View more View less

          Gene Aliases: 2310021G14Rik; B130064M09Rik; C25KG; RAB27B

          View more View less

          UniProt ID: (Human) O00194, (Mouse) Q99P58, (Rat) Q99P74

          View more View less

          Entrez Gene ID: (Human) 5874, (Mouse) 80718, (Rat) 84590

          View more View less

          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding hydrolase activity GDP binding protein domain specific binding myosin V binding
          Process(es)
          exocytosis regulation of exocytosis positive regulation of exocytosis synaptic vesicle endocytosis multivesicular body sorting pathway anterograde axonal protein transport intracellular protein transport vesicle docking involved in exocytosis metabolic process protein secretion melanosome transport Rab protein signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-wk68q:80/100.66.79.78:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline