Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • DENND5A Antibodies

          Abnova

          RAB6IP1 Polyclonal Antibody, MaxPab™

          View all (4) DENND5A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAB6IP1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RAB6IP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAB6IP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAB6IP1 Antibody (H00023258-B01P) in WB

          Western Blot analysis of DENND5A expression in transfected 293T cell line by DENND5A MaxPab polyclonal antibody. Lane 1: RAB6IP1 transfected lysate(141.57 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAB6IP1 Antibody in Western Blot (WB)

          Product Details

          H00023258-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAB6IP1 (AAH71596.1, 1 a.a. approximately 1287 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSGGGGGGGS APSRFADYFV ICGLDTETGL EPDELSALCQ YIQASKARDG ASPFISSTTE GENFEQTPLR RTFKSKVLAR YPENVEWNPF DQDAVGMLCM PKGLAFKTQA DPREPQFHAF IITREDGSRT FGFALTFYEE VTSKQICSAM QTLYHMHNAE YDVLHAPPAD DRDQSSMEDG EDTPVTKLQR FNSYDISRDT LYVSKCICLI TPMSFMKACR SVLQQLHQAV TSPQPPPLPL ESYIYNVLYE VPLPPPGRSL KFSGVYGPII CQRPSTNELP LFDFPVKEVF ELLGVENVFQ LFTCALLEFQ ILLYSQHYQR LMTVAETITA LMFPFQWQHV YVPILPASLL HFLDAPVPYL MGLHSNGLDD RSKLELPQEA NLCFVDIDNH FIELPEDLPQ FPNKLEFVQE VSEILMAFGI PPEGNLHCSE SASKLKRLRA SELVSDKRNG NIAGSPLHSY ELLKENETIA RLQALVKRTG VSLEKLEVRE DPSSNKDLKV QCDEEELRIY QLNIQIREVF ANRFTQMFAD YEVFVIQPSQ DKESWFTNRE QMQNFDKASF LSDQPEPYLP FLSRFLETQM FASFIDNKIM CHDDDDKDPV LRVFDSRVDK IRLLNVRTPT LRTSMYQKCT TVDEAEKAIE LRLAKIDHTA IHPHLLDMKI GQGKYEPGFF PKLQSDVLST GPASNKWTKR NAPAQWRRKD RQKQHTEHLR LDNDQREKYI QEARTMGSTI RQPKLSNLSP SVIAQTNWKF VEGLLKECRN KTKRMLVEKM GREAVELGHG EVNITGVEEN TLIASLCDLL ERIWSHGLQV KQGKSALWSH LLHYQDNRQR KLTSGSLSTS GILLDSERRK SDASSLMPPL RISLIQDMRH IQNIGEIKTD VGKARAWVRL SMEKKLLSRH LKQLLSDHEL TKKLYKRYAF LRCDDEKEQF LYHLLSFNAV DYFCFTNVFT TILIPYHILI VPSKKLGGSM FTANPWICIS GELGETQIMQ IPRNVLEMTF ECQNLGKLTT VQIGHDNSGL YAKWLVEYVM VRNEITGHTY KFPCGRWLGK GMDDGSLERI LVGELLTSQP EVDERPCRTP PLQQSPSVIR RLVTISPNNK PKLNTGQIQE SIGEAVNGIV KHFHKPEKER GSLTLLLCGE CGLVSALEQA FQHGFKSPRL FKNVFIWDFL EKAQTYYETL EKNEVVPEEN WHTRARNFCR FVTAINNTPR NIGKDGKFQM LVCLGARDHL LHHWIALLAD CPITAHMYED VALIKDHTLV NSLIRVLQTL QEFNITLETS LVKGIDI

          Target Information

          DENND5A (DENN domain-containing protein 5A) is a guanine nucleotide exchange factor (GEF) which activates RAB6A and RAB39A/RAB39B. It promotes the exchange of GDP to GTP by converting inactive GDP-bound Rab proteins to their active GTP form. DENND5A is also involved in the negative regulation of neurtie outgrowth. Mutations affecting this gene can result in epileptic encephalopathy, early infantile, 49 (EIEE49).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DENN domain-containing protein 5A; DENN/MADD domain containing 5A; RAB6 interacting protein 1; Rab6-interacting protein 1; Rab6IP1; unnamed protein product

          View more View less

          Gene Aliases: DEE49; DENND5A; EIEE49; KIAA1091; RAB6IP1

          View more View less

          UniProt ID: (Human) Q6IQ26

          View more View less

          Entrez Gene ID: (Human) 23258

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding small GTPase binding
          Process(es)
          negative regulation of neuron projection development retrograde transport, endosome to Golgi
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline