Search
Search
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: RA9B.
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-associated protein 9B; Ras-associated protein RAB9-like; TRAB9; unnamed protein product
Gene Aliases: 9330195C02Rik; Rab-9L; RAB9L; RGD1562951
UniProt ID: (Human) Q9NP90, (Mouse) Q8BHH2
Entrez Gene ID: (Human) 51209, (Rat) 367915, (Mouse) 319642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support