Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RABL3 Antibodies

          Proteintech

          RABL3 Polyclonal Antibody

          2 References
          View all (5) RABL3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RABL3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          RABL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RABL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RABL3 Antibody (11916-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed A549 cells using RABL3 antibody (Product # 11916-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RABL3 Antibody in Immunocytochemistry (ICC/IF)
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RABL3 Antibody in Western Blot (WB)
          RABL3 Antibody in Western Blot (WB)
          RABL3 Antibody in Western Blot (WB)
          RABL3 Antibody in Immunoprecipitation (IP)
          RABL3 Polyclonal Antibody

          Product Details

          11916-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:400
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RABL3 Fusion Protein Ag2531 (1-236 aa encoded by BC020832)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MASLDRVKV LVLGDSGVGK SSLVHLLCQN QVLGNPSWTV GCSVDVRVHD YKEGTPEEKT CYIELWDVGG SVGSASSVKS TRAVFYNSVN GIIFVHDLTN KKSSQNLRRW SLEALNRDLV PTGVLVTNGD YDQEQFADNQ IPLLVIGTKL DQIHETKRHE VLTTTAFLAE DFNPEEINLD CTNPRYLAAG SSNAVKLSRF FDKVIEKRYF LREGNQIPGF PDRKRFGAGT LKSLHYD (1-236 aa encoded by BC020832)

          Target Information

          Predicted to enable GTP binding activity; GTPase activity; and protein homodimerization activity. Involved in regulation of Ras protein signal transduction and regulation of protein lipidation. Predicted to be active in endomembrane system. Implicated in pancreatic cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: RAB, member of RAS oncogene family-like 3; Rab-like protein 3; unnamed protein product

          View more View less

          Gene Aliases: 4930553C05Rik; AA958834; PNCA5; RABL3

          View more View less

          UniProt ID: (Human) Q5HYI8, (Mouse) Q9D4V7

          View more View less

          Entrez Gene ID: (Human) 285282, (Mouse) 67657

          View more View less

          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding protein homodimerization activity molecular_function
          Process(es)
          natural killer cell differentiation intracellular protein transport B cell differentiation T cell differentiation in thymus regulation of Ras protein signal transduction protein stabilization regulation of protein lipidation small GTPase mediated signal transduction biological_process Rab protein signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline