Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAG2 Antibodies

          Abnova

          RAG2 Polyclonal Antibody, MaxPab™

          View all (18) RAG2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAG2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RAG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAG2 Antibody (H00005897-D01P) in WB

          Western Blot analysis of RAG2 expression in transfected 293T cell line by RAG2 MaxPab polyclonal antibody. Lane 1: RAG2 transfected lysate(59.20 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAG2 Antibody in Western Blot (WB)

          Product Details

          H00005897-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAG2 (NP_000527.1, 1 a.a. approximately 527 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSLQMVTVSN NIALIQPGFS LMNFDGQVFF FGQKGWPKRS CPTGVFHLDV KHNHVKLKPT IFSKDSCYLP PLRYPATCTF KGSLESEKHQ YIIHGGKTPN NEVSDKIYVM SIVCKNNKKV TFRCTEKDLV GDVPEARYGH SINVVYSRGK SMGALFGGRS YMPSTHRTTE KWNSVADCLP CVFLVDFEFG CATSYILPEL QDGLSFHVSI AKNDTIYILG GHSLANNIRP ANLYRIRVDL PLGSPAVNCT VLPGGISVSS AILTQTNNDE FVIVGGYQLE NQKRMICNII SLEDNKIEIR EMETPDWTPD IKHSKIWFGS NTGNGTVFLG IPGDNKQVVS EGFYFYMLKC AEDDTNEEQT TFTNSQTSTE DPGDSTPFED SEEFCFSAEA NSFDGDDEFD TYNEDDEEDE SETGYWITCC PTCDVDINTW VPFYSTELNK PAMIYCSHGD GHWVHAQCMD LAERTLIHLS AGSNKYYCNE HVEIARALHT PQRVLPLKKP PMKSLRKKGS GKILTPAKKS FLRRLFD

          Target Information

          RAG2 (V(D)J recombination-activating protein 2) is a core component of the RAG complex - a multiprotein complex that mediates the DNA cleavage phase during V(D)J recombination. V(D)J recombination assembles a diverse repertoire of immunoglobulin and T-cell receptor genes in developing B and T-lymphocytes through rearrangement of different V (variable), in some cases D (diversity) and J (joining) gene segments. DNA cleavage by the RAG complex occurs in 2 steps: 1) first a nick is introduced in the top strand immediately upstream of the heptamer, generating a 3'-hydroxyl group that can attack the phosphodiester bond on the opposite strand in a direct transesterification reaction, thereby creating 4 DNA ends. 2) hairpin coding ends and 2 blunt, 5'-phosphorylated ends are created. The chromatin structure plays an essential rold in the V(D)J recombination reactions and the presence of histone H3 trimethylated at 'Lys-4' (H3K4me3) stimulates both the nicking and hair pinning expressed on individual B-lymphocytes. The introduction of DNA breaks by the RAG complex on one immunoglobulin allele induces ATM-dependent repositioning of the other allele to pericentromeric heterochromatin. This prevents accessibility to the RAG complex and recombination of the second allele. In the RAG complex, RAG2 is not the catalytic component but is required for all known catalytic activities mediated by RAG1. Mutations affecting the gene can results in combined cellular and humoral immune defects with granulomas, severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-negative/NK-cell-positive, and Omenn syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: lymphocyte-specific component of V(D)J recombinase; RAG 2; RAG-2; recombination activating gene 2; recombination activating protein 2; unnamed protein product; V(D)J recombination-activating protein 2

          View more View less

          Gene Aliases: RAG-2; RAG2

          View more View less

          UniProt ID: (Human) P55895

          View more View less

          Entrez Gene ID: (Human) 5897, (Rat) 295953

          View more View less

          Function(s)
          DNA binding chromatin binding phosphatidylinositol-4,5-bisphosphate binding phosphatidylinositol-3,4,5-trisphosphate binding zinc ion binding phosphatidylinositol binding phosphatidylinositol-3,4-bisphosphate binding sequence-specific DNA binding metal ion binding phosphatidylinositol-3,5-bisphosphate binding histone H3K4me3 reader activity molecular_function methylated histone binding
          Process(es)
          mature B cell differentiation involved in immune response B cell lineage commitment pre-B cell allelic exclusion B cell homeostatic proliferation T cell lineage commitment DNA recombination chromatin organization B cell differentiation T cell differentiation T cell differentiation in thymus negative regulation of T cell differentiation in thymus V(D)J recombination organ growth defense response to bacterium positive regulation of organ growth DN2 thymocyte differentiation biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0