Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RAP1A/RAP1B Antibodies

          Invitrogen

          RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (6C7G0)

          View all (5) RAP1A/RAP1B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (6C7G0)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAP1A/RAP1B Antibody (MA5-38205) in ICC/IF

          Immunofluorescence analysis of RAP1A/RAP1B in NIH-3T3 cells. Samples were incubated with RAP1A/RAP1B Monoclonal antibody (Product # MA5-38205) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          RAP1A/RAP1B Antibody in Immunocytochemistry (ICC/IF)
          RAP1A/RAP1B Antibody in Western Blot (WB)
          RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (6C7G0)

          Product Details

          MA5-38205

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          6C7G0

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 70-150 of human RAP1A + RAP1B (P61224)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          40 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2898122

          Product Specific Information

          Positive Samples: HeLa, NIH/3T3, C6, U-251MG, HUVEC, Rat lung, Rat spleen

          Immunogen sequence: LYMKNGQGFA LVYSITAQST FNDLQDLREQ ILRVKDTDDV PMILVGNKCD LEDERVVGKE QGQNLARQWN NCAFLESSAK S

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; NB4 apoptosis/differentiation related protein; RAP1B; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; Ras-related protein Rap-1b; RAS-related protein RAP1B; small GTP binding protein; unnamed protein product

          View more View less

          Gene Aliases: 2810443E11Rik; AI848598; C21KG; G-22K; K-REV; KREV-1; KREV1; OK/SW-cl.11; RAL1B; rap 1A; RAP-1A; RAP1; RAP1A; RAP1B; SMGP21; THC11

          View more View less

          UniProt ID: (Human) P62834, (Human) P61224, (Mouse) P62835, (Rat) P62836, (Rat) Q62636, (Mouse) Q99JI6

          View more View less

          Entrez Gene ID: (Human) 5906, (Human) 5908, (Mouse) 109905, (Rat) 295347, (Rat) 171337, (Mouse) 215449

          View more View less

          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity guanyl-nucleotide exchange factor activity protein binding GTP binding hydrolase activity GDP binding small GTPase binding macromolecular complex binding protein transporter activity Ras GTPase binding Rap guanyl-nucleotide exchange factor activity protein complex binding small GTPase
          Process(es)
          signal transduction small GTPase mediated signal transduction nervous system development response to carbohydrate positive regulation of neuron projection development synaptic vesicle exocytosis Rap protein signal transduction negative regulation of collagen biosynthetic process nerve growth factor signaling pathway positive regulation of GTPase activity positive regulation of protein kinase activity positive regulation of glucose import positive regulation of phagocytosis positive regulation of Fc receptor mediated stimulatory signaling pathway establishment of endothelial barrier positive regulation of ERK1 and ERK2 cascade cellular response to cAMP protein localization to plasma membrane response to antineoplastic agent liver regeneration regulation of neurotransmitter receptor localization to postsynaptic specialization membrane regulation of cell junction assembly cellular response to forskolin cellular response to nerve growth factor stimulus negative regulation of synaptic vesicle exocytosis positive regulation of vasculogenesis cell proliferation calcium ion regulated exocytosis positive regulation of integrin activation negative regulation of calcium ion-dependent exocytosis establishment of localization in cell modification of postsynaptic structure regulation of establishment of cell polarity protein transport metabolic process microvillus assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline