Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RAPGEF3 Antibodies

          Invitrogen

          RAPGEF3 Polyclonal Antibody

          View all (23) RAPGEF3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAPGEF3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RAPGEF3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAPGEF3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAPGEF3 Antibody (PA5-102603) in WB

          Western blot analysis of RAPGEF3 in HepG2 cell lysate (left lane: treated with the antigen-specific peptide). Samples were incubated with RAPGEF3 polyclonal antibody (Product # PA5-102603). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAPGEF3 Antibody in Western Blot (WB)
          RAPGEF3 Polyclonal Antibody

          Product Details

          PA5-102603

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human RAPGEF3(Accession O95398), corresponding to amino acid residues S753-A803.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2852005

          Product Specific Information

          Antibody detects endogenous levels of total RAPGEF3.

          Target Information

          Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 9330170P05Rik; cAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); EPAC 1; Exchange factor directly activated by cAMP 1; Exchange protein directly activated by cAMP 1; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; unnamed protein product

          View more View less

          Gene Aliases: 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; CGEF1; EPAC; EPAC1; HSU79275; RAPGEF3

          View more View less

          UniProt ID: (Human) O95398, (Rat) Q9Z1C8, (Mouse) Q8VCC8

          View more View less

          Entrez Gene ID: (Human) 10411, (Rat) 59326, (Mouse) 223864

          View more View less

          Function(s)
          nucleotide binding guanyl-nucleotide exchange factor activity protein binding protein domain specific binding cAMP binding cAMP-dependent protein kinase regulator activity Rap guanyl-nucleotide exchange factor activity ion channel binding guanyl-nucleotide exchange factor
          Process(es)
          angiogenesis adaptive immune response signal transduction adenylate cyclase-activating G-protein coupled receptor signaling pathway small GTPase mediated signal transduction Ras protein signal transduction associative learning Rap protein signal transduction regulation of actin cytoskeleton organization negative regulation of syncytium formation by plasma membrane fusion intracellular signal transduction protein kinase B signaling positive regulation of GTPase activity regulation of angiogenesis positive regulation of angiogenesis positive regulation of protein export from nucleus regulation of insulin secretion positive regulation of stress fiber assembly regulation of protein kinase B signaling positive regulation of syncytium formation by plasma membrane fusion establishment of endothelial barrier cellular response to cAMP positive regulation of peptidyl-serine phosphorylation regulation of protein kinase activity positive regulation of protein acetylation regulation of actin cytoskeleton reorganization regulation of histone H3-K9 acetylation regulation of protein phosphorylation positive regulation of protein phosphorylation positive regulation of smooth muscle cell migration positive regulation of ATPase activity negative regulation of collagen biosynthetic process positive regulation of cell size positive regulation of transcription from RNA polymerase II promoter positive regulation of binding positive regulation of calcium ion transport negative regulation of cell death positive regulation of actin cytoskeleton reorganization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline