Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RAPGEF5 Antibodies

          Proteintech

          RAPGEF5 Polyclonal Antibody

          2 References
          View all (6) RAPGEF5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAPGEF5 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          RAPGEF5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAPGEF5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAPGEF5 Antibody (12556-1-AP) in WB

          Mouse brain tissue were subjected to SDS PAGE followed by western blot with 12556-1-AP (RAPGEF5 antibody) at dilution of 1:600 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Antibody in Western Blot (WB)
          RAPGEF5 Polyclonal Antibody

          Product Details

          12556-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAPGEF5 Fusion Protein Ag3229 (129-444 aa encoded by BC039203)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          37.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: KKYQGKEEN SDVPRRKRKV LHLVSQWIAL YKDWLPEDEH SKMFLKTIYR NVLDDVYEYP ILEKELKEFQ KILGMHRRHT VDEYSPQKKN KALFHQFSLK ENWLQHRGTV TETEEIFCHV YITEHSYVSV KAKVSSIAQE ILLCSQLGKR VQLVKKFIKI AAHCKAQRNL NSFFAIVMGL NTASVSRLSQ TWEKIPGKFK KLFSELESLT DPSLNHKAYR DAFKKMKPPK IPFVPLLLKD VTFIHEGNKT FLDNLVNFEK LHMIADTVRT LRHCRTNQFG DLSPKEHQEL KSYVNHLYVI DSQQALFELS HRIEPRV (129-444 aa encoded by BC039203 )

          Target Information

          Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as Repac, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation. Repac, also known as Rap guanine nucleotide exchange factor 5 (RAPGEF5), is a 580 amino acid protein that is expressed in brain and testis, with weaker expression in heart, placenta, lung, pancreas and small intestine. Existing as two alternatively spliced isoforms, the Repac gene is conserved in cow, mouse and rat. The gene that encodes Repac maps to human chromosome 7p15.3, with high expression of this region linking to positive response to erlotinib therapy for those with non-small-cell lung cancer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Guanine nucleotide exchange factor for Rap1; KIAA0277; M-Ras-regulated GEF; M-Ras-regulated Rap GEF; MR-GEF; Mras regulated guanine nucleotide exchange factor; Rap guanine nucleotide exchange factor (GEF) 5; Rap guanine nucleotide exchange factor (GEF) 5 isoform RAPGEF5_1; Rap guanine nucleotide exchange factor (GEF) 5 isoform RAPGEF5_2; Rap guanine nucleotide exchange factor 5; Related to Epac; Repac; tcag7.1034; unnamed protein product

          View more View less

          Gene Aliases: 4932413M22; C86120; D030051B22Rik; GFR; KIAA0277; mKIAA0277; mmr-gef; MR-GEF; MRGEF; RAPGEF5; REPAC

          View more View less

          UniProt ID: (Human) Q92565, (Mouse) Q8C0Q9

          View more View less

          Entrez Gene ID: (Human) 9771, (Mouse) 217944

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding GTP-dependent protein binding Rap guanyl-nucleotide exchange factor activity guanyl-nucleotide exchange factor
          Process(es)
          small GTPase mediated signal transduction Ras protein signal transduction nervous system development intracellular signal transduction positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0