Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RARB Antibodies

          Proteintech

          RARB Polyclonal Antibody

          6 References
          View all (13) RARB antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RARB Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          RARB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RARB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RARB Antibody (14013-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using RARB antibody (14013-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L), CL594-phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RARB Antibody in Immunocytochemistry (ICC/IF)
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RARB Antibody in Western Blot (WB)
          RARB Polyclonal Antibody

          Product Details

          14013-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RARB fusion protein Ag5119
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.6 mg/mL

          Amount

          90 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: EGCKGFFRRS IQKNMIYTCH RDKNCVINKV TRNRCQYCRL QKCFEVGMSK ESVRNDRNKK KKETSKQECT ESYEMTAELD DLTEKIRKAH QETFPSLCQL GKYTTNSSAD HRVRLDLGLW DKFSELATKC IIKIVEFAKR LPGFTGLTIA DQITLLKAAC LDILILRICT RYTPEQDTMT FSDGLTLNRT QMHNAGFGPL TDLVFTFANQ LLPLEMDDTE TGLLSAICLI CGDRQDLEEP TKVDKLQEPL LEALKIYIRK RRPSKPHMFP KILMKITDLR SISAKGAERV ITLKMEIPGS MPPLIQEMLE NSEGHEPLTP SSSGNTAEHS PSISPSSVEN SGVSQSPLVQ

          Target Information

          Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HBV-activated protein; hepatitis B virus activated protein; Nuclear receptor subfamily 1 group B member 2; nuclear receptor; RAR beta; open reading frame 1; putative; RAR beta; RAR beta 1'; RAR-b; RAR-beta; RAR-Beta2; RAR-Beta4; RAR-epsilon; RARB; RARbeta isoform 5; retinoic acid; retinoic acid receptor; Retinoic acid receptor beta; retinoic acid receptor, beta polypeptide; unnamed protein product

          View more View less

          Gene Aliases: A830025K23; HAP; MCOPS12; NR1B2; RARB; RARbeta; RARbeta1; RRB2

          View more View less

          UniProt ID: (Human) P10826, (Mouse) P22605

          View more View less

          Entrez Gene ID: (Human) 5915, (Mouse) 218772

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding sequence-specific DNA binding macromolecular complex binding metal ion binding retinoid X receptor binding heterocyclic compound binding sequence-specific double-stranded DNA binding RNA polymerase II regulatory region sequence-specific DNA binding steroid hormone receptor activity retinoic acid receptor activity drug binding protein complex binding C4 zinc finger nuclear receptor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ureteric bud development glandular epithelial cell development outflow tract septum morphogenesis growth plate cartilage development regulation of transcription, DNA-templated apoptotic process signal transduction cell proliferation programmed cell death striatum development neurogenesis cell differentiation regulation of myelination negative regulation of chondrocyte differentiation embryonic hindlimb morphogenesis multicellular organism growth positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of programmed cell death positive regulation of transcription from RNA polymerase II promoter embryonic eye morphogenesis retinoic acid receptor signaling pathway embryonic digestive tract development ventricular cardiac muscle cell differentiation bone development negative regulation of cartilage development neural precursor cell proliferation stem cell proliferation negative regulation of stem cell proliferation retinal pigment epithelium development transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter positive regulation of cell proliferation negative regulation of cell proliferation steroid hormone mediated signaling pathway positive regulation of neuron differentiation retina development in camera-type eye
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline