Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MMAAEAGSE EGGPVTAGAG GGGAAAGSSA YPAVCRVKIP AALPVAAAPY PGLVETGVAG TLGGGAALGS EFLGAGSVAG ALGGAGLTGG GTAAGVAGAA AGVAGAAVAG PSGDMALTKL PTSLLAETLG PGGGFPPLPP PPYLPPLGAG LGTVDEGDSL DGPEYEEEEV AIPLTAPPTN QWYHGKLDRT IAEERLRQAG KSGSYLIRES DRRPGSFVLS FLSQMNVVNH FRIIAMCGDY YIGGRRFSSL SDLIGYYSHV SCLLKGEKLL YPVAPPEPVE DRRRVRAILP YTKVPDTDEI SFLKGDMFIV HNELEDGWMW VTNLRTDEQG L
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAP; GTPase-activating protein; p120 RAS GTPase activating protein; p120-rasGAP; p120GAP; Ras GTPase-activating protein 1; Ras p21 protein activator; RAS p21 protein activator (GTPase activating protein) 1; RAS p21 protein activator 1; rasGAP; triphosphatase-activating protein; unnamed protein product
Gene Aliases: CM-AVM; CMAVM; CMAVM1; GAP; GAPX; p120; p120GAP; p120RASGAP; PKWS; RASA; RASA1; RASGAP
UniProt ID: (Human) P20936, (Rat) P50904
Entrez Gene ID: (Human) 5921, (Rat) 25676, (Mouse) 218397
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support