Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RASGRP4 Antibodies

          Abnova

          RASGRP4 Polyclonal Antibody, MaxPab™

          View all (5) RASGRP4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RASGRP4 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          RASGRP4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RASGRP4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RASGRP4 Antibody (H00115727-D01P) in WB

          Western Blot analysis of RASGRP4 expression in transfected 293T cell line (H00115727-T01) by RASGRP4 MaxPab polyclonal antibody.

          Lane 1: RASGRP4 transfected lysate(74.90 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RASGRP4 Antibody in Western Blot (WB)
          RASGRP4 Antibody in Proximity Ligation Assay (PLA) (PLA)
          RASGRP4 Polyclonal Antibody, MaxPab™

          Product Details

          H00115727-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RASGRP4 (NP_733749.1, 1 a.a. approximately 673 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MNRKDSKRKS HQECTGKIGG RGRPRQVRRH KTCPSPREIS KVMASMNLGL LSEGGCSEDE LLEKCIQSFD SAGSLCHEDH MLNMVLAMHS WVLPSADLAA RLLTSYQKAT GDTQELRRLQ ICHLVRYWLM RHPEVMHQDP QLEEVIGRFW ATVAREGNSA QRRLGDSSDL LSPGGPGPPL PMSSPGLGKK RKVSLLFDHL ETGELAQHLT YLEFRSFQAI TPQDLRSYVL QGSVRGCPAL EGSVGLSNSV SRWVQVMVLS RPGPLQRAQV LDKFIHVAQR LHQLQNFNTL MAVTGGLCHS AISRLKDSHA HLSPDSTKAL LELTELLASH NNYARYRRTW AGCAGFRLPV LGVHLKDLVS LHEAQPDRLP DGRLHLPKLN NLYLRLQELV ALQGQHPPCS ANEDLLHLLT LSLDLFYTED EIYELSYARE PRCPKSLPPS PFNAPLVVEW APGVTPKPDR VTLGRHVEQL VESVFKNYDP EGRGTISQED FERLSGNFPF ACHGLHPPPR QGRGSFSREE LTGYLLRASA ICSKLGLAFL HTFHEVTFRK PTFCDSCSGF LWGVTKQGYR CRECGLCCHK HCRDQVKVEC KKRPGAKGDA GPPGAPVPST PAPHASCGSE ENHSYTLSLE PETGCQLRHA WTQTESPHPS WETDTVPCPV MDPPSTASSK LDS

          Target Information

          The RASGRP4 (RAS Guanyl Releasing Protein 4) gene encodes a guanine nucleotide exchange factor (GEF) that specifically activates RAS proteins by facilitating the exchange of GDP for GTP. Located on chromosome 19, RASGRP4 is expressed primarily in hematopoietic cells, including mast cells and certain myeloid cells. The protein contains a diacylglycerol (DAG)-binding C1 domain, a Ras exchange motif, and a CDC25 domain, which are essential for its activation and function. RASGRP4 plays a pivotal role in signal transduction pathways that regulate cell proliferation, differentiation, and survival. Dysregulation of RASGRP4 has been linked to various diseases, including inflammatory disorders and cancers, particularly those involving the hematopoietic system. Studies have shown that overexpression or mutations in RASGRP4 can lead to aberrant RAS signaling, contributing to oncogenesis and disease progression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: guanyl nucleotide releasing protein 4; hRasGRP4; hRasGRP4-v1; hRasGRP4-v2; RAS guanyl-releasing protein 4; RasGRP4

          View more View less

          Gene Aliases: RASGRP4

          View more View less

          UniProt ID: (Human) Q8TDF6

          View more View less

          Entrez Gene ID: (Human) 115727

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity calcium ion binding protein binding zinc ion binding lipid binding diacylglycerol binding GTPase regulator activity metal ion binding
          Process(es)
          positive regulation of natural killer cell mediated immunity phospholipase C-activating G-protein coupled receptor signaling pathway small GTPase mediated signal transduction Ras protein signal transduction myeloid cell differentiation cell differentiation antibacterial innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline