Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (81%), Pig (96%), Rhesus Monkey (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The function of this gene has not yet been determined but may involve a role in tumor suppression. Alternative splicing of this gene results in several transcript variants; however, most of the variants have not been fully described.