Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RBM8A Antibodies

          Proteintech

          RBM8A/Y14 Polyclonal Antibody

          5 References
          View all (16) RBM8A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RBM8A/Y14 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (15)
          RBM8A/Y14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RBM8A/Y14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RBM8A/Y14 Antibody (14958-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HeLa cells using 14958-1-AP (RBM8A,Y14 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RBM8A/Y14 Antibody in Immunocytochemistry (ICC/IF)
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RBM8A/Y14 Antibody in Western Blot (WB)
          RBM8A/Y14 Antibody in Western Blot (WB)
          RBM8A/Y14 Antibody in Western Blot (WB)
          RBM8A/Y14 Antibody in Western Blot (WB)
          RBM8A/Y14 Antibody in Western Blot (WB)
          RBM8A/Y14 Antibody in Immunoprecipitation (IP)
          RBM8A/Y14 Polyclonal Antibody

          Product Details

          14958-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RBM8A,Y14 Fusion Protein Ag6800 (1-174 aa encoded by BC017088)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          33 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MADVLDLHE AGGEDFAMDE DGDESIHKLK EKAKKRKGRG FGSEEGSRAR MREDYDSVEQ DGDEPGPQRS VEGWILFVTG VHEEATEEDI HDKFAEYGEI KNIHLNLDRR TGYLKGYTLV EYETYKEAQA AMEGLNGQDL MGQPISVDWC FVRGPPKGKR RGGRRRSRSP DRRRR (1-174 aa encoded by BC017088)

          Target Information

          This gene encodes a protein with a conserved RNA-binding motif. The protein is found predominantly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: binder of ovarian cancer 1 protein BOV-1; binder of OVCA1; Binder of OVCA1-1; BOV-1; BOV1; BOV1A; BOV1B; BOV1C; exon junction complex core component Y14; RBM8A; ribonucleoprotein RBM8; Ribonucleoprotein RBM8A; RNA binding motif protein 8; RNA binding motif protein 8B; rna-binding; RNA-binding motif protein 8A; RNA-binding protein 8A; RNA-binding protein Y14; unnamed protein product

          View more View less

          Gene Aliases: 2310057C03Rik; AA673428; BOV-1A; BOV-1B; BOV-1C; C1DELq21.1; DEL1q21.1; HSPC114; MDS014; RBM8; RBM8A; RBM8B; TAR; Y14; ZNRP; ZRNP1

          View more View less

          UniProt ID: (Human) Q9Y5S9, (Mouse) Q9CWZ3, (Rat) Q27W01

          View more View less

          Entrez Gene ID: (Human) 9939, (Mouse) 60365, (Rat) 295284

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding protein binding poly(A) RNA binding nucleotide binding
          Process(es)
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay regulation of alternative mRNA splicing, via spliceosome mRNA splicing, via spliceosome RNA processing mRNA processing mRNA export from nucleus regulation of translation RNA splicing regulation of mRNA processing mRNA transport regulation of nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0