Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • RECQL5 Antibodies

          Proteintech

          RECQL5 Polyclonal Antibody

          View all (16) RECQL5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RECQL5 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          RECQL5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RECQL5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RECQL5 Antibody (12468-2-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using RECQL5 antibody 12468-2-AP at 1:100 dilution and FITC-labeled donkey anti-rabbit IGG (green). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RECQL5 Antibody in Immunocytochemistry (ICC/IF)
          RECQL5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)
          RECQL5 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RECQL5 Polyclonal Antibody

          Product Details

          12468-2-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RECQL5 Fusion Protein Ag3145 (1-350 aa encoded by BC016911)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSSHHTTFP FDPERRVRST LKKVFGFDSF KTPLQESATM AVVKGNKDVF VCMPTGAGKS LCYQLPALLA KGITIVVSPL IALIQDQVDH LLTLKVRVSS LNSKLSAQER KELLADLERE KPQTKILYIT PEMAASSSFQ PTLNSLVSRH LLSYLVVDEA HCVSQWGHDF RPDYLRLGAL RSRLGHAPCV ALTATATPQV QEDVFAALHL KKPVAIFKTP CFRANLFYDV QFKELISDPY GNLKDFCLKA LGQEADKGLS GCGIVYCRTR EACEQLAIEL SCRGVNAKAY HAGLKASERT LVQNDWMEEK VPVIVATISF GMGVDKANVR FVAHWNIAKS MAGYYQESGR A (1-350 aa encoded by BC016911)

          Target Information

          RECQL5 gene ontology annotations related to this gene include cellular response to camptothecin; chromosome separation; DNA duplex unwinding; DNA recombination; DNA repair; DNA replication; double-strand break repair via homologous recombination; mitotic sister chromatid segregation; negative regulation of double-strand break repair via homologous recombination; negative regulation of mitotic cell cycle; negative regulation of transcription elongation from RNA polymerase II promoter; replication-born double-strand break repair via sister chromatid exchange; response to X-ray.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DNA 3'-5' helicase RecQ5; DNA helicase, RecQ-like type 5; FLJ90603; RecQ helicase-like 5; RecQ protein 5; RecQ protein-like 5; RecQ5; RECQL5; RECQL5beta; unnamed protein product

          View more View less

          Gene Aliases: RECQ5; Recq5b; Recql5b

          View more View less

          UniProt ID: (Human) O94762, (Mouse) Q8VID5, (Rat) D4ACP5

          View more View less

          Entrez Gene ID: (Human) 9400, (Mouse) 170472, (Rat) 287834

          View more View less

          Function(s)
          RNA polymerase II core binding nucleic acid binding DNA helicase activity helicase activity protein binding ATP binding four-way junction helicase activity ATPase activity identical protein binding 3'-5' DNA helicase activity catalytic activity, acting on DNA
          Process(es)
          mitotic cell cycle double-strand break repair via homologous recombination DNA replication DNA repair DNA recombination regulation of transcription, DNA-templated negative regulation of transcription elongation from RNA polymerase II promoter chromosome separation cellular response to xenobiotic stimulus cellular response to camptothecin replication-born double-strand break repair via sister chromatid exchange mitotic DNA-dependent DNA replication negative regulation of double-strand break repair via homologous recombination DNA metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0