Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Regucalcin Antibodies

          Proteintech

          RGN/SMP30 Polyclonal Antibody

          10 References
          View all (8) Regucalcin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RGN/SMP30 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RGN/SMP30 Antibody (17947-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded rat kidney tissue slide using (Product # 17947-1-AP) (RGN/SMP30 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RGN/SMP30 Antibody in Western Blot (WB)
          RGN/SMP30 Antibody in Western Blot (WB)
          RGN/SMP30 Antibody in Immunoprecipitation (IP)
          RGN/SMP30 Polyclonal Antibody

          Product Details

          17947-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:3000-1:10,000
          View 9 publications 9 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Bovine, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RGN/SMP30 Fusion Protein Ag12423 (1-299 aa encoded by BC073173)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSSIKIECV LPENCRCGES PVWEEVSNSL LFVDIPAKKV CRWDSFTKQV QRVTMDAPVS SVALRQSGGY VATIGTKFCA LNWKEQSAVV LATVDNDKKN NRFNDGKVDP AGRYFAGTMA EETAPAVLER HQGALYSLFP DHHVKKYFDQ VDISNGLDWS LDHKIFYYID SLSYSVDAFD YDLQTGQISN RRSVYKLEKE EQIPDGMCID AEGKLWVACY NGGRVIRLDP VTGKRLQTVK LPVDKTTSCC FGGKNYSEMY VTCARDGMDP EGLLRQPEAG GIFKITGLGV KGIAPYSYAG (1-299 aa encoded by BC073173)

          Target Information

          The protein encoded by this gene is a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5' UTRs, but encoding the same protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis secretory protein Li 41; Gluconolactonase; GNL; RC; Regucalcin; Senescence marker protein 30; SMP-30; unnamed protein product

          View more View less

          Gene Aliases: AI265316; GNL; HEL-S-41; RC; Reguc; RGN; SMP30

          View more View less

          UniProt ID: (Human) Q15493, (Rat) Q03336, (Mouse) Q64374

          View more View less

          Entrez Gene ID: (Human) 9104, (Rat) 25106, (Mouse) 19733

          View more View less

          Function(s)
          gluconolactonase activity calcium ion binding zinc ion binding hydrolase activity enzyme regulator activity metal ion binding esterase
          Process(es)
          cellular calcium ion homeostasis positive regulation of ATPase activity regulation of calcium-mediated signaling kidney development liver development negative regulation of protein phosphorylation negative regulation of protein kinase activity spermatogenesis aging positive regulation of triglyceride biosynthetic process positive regulation of glucose metabolic process positive regulation of phosphatase activity L-ascorbic acid biosynthetic process negative regulation of phosphoprotein phosphatase activity negative regulation of GTPase activity negative regulation of apoptotic process positive regulation of GTPase activity negative regulation of nitric oxide biosynthetic process positive regulation of fatty acid biosynthetic process negative regulation of epithelial cell proliferation negative regulation of cyclic-nucleotide phosphodiesterase activity liver regeneration negative regulation of sperm motility positive regulation of superoxide dismutase activity positive regulation of calcium-transporting ATPase activity negative regulation of RNA biosynthetic process negative regulation of bone development positive regulation of proteolysis involved in cellular protein catabolic process negative regulation of calcium-dependent ATPase activity negative regulation of DNA catabolic process positive regulation of dUTP diphosphatase activity negative regulation of leucine-tRNA ligase activity negative regulation of DNA biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline