Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RGS9 Antibodies

          Proteintech

          RGS9 Polyclonal Antibody

          1 Reference
          View all (5) RGS9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RGS9 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          RGS9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RGS9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RGS9 Antibody (17970-1-AP) in WB

          Mouse brain tissue were subjected to SDS PAGE followed by western blot with 17970-1-AP (RGS9 antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RGS9 Antibody in Western Blot (WB)
          RGS9 Antibody in Western Blot (WB)
          RGS9 Polyclonal Antibody

          Product Details

          17970-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RGS9 Fusion Protein Ag11976 (273-622 aa encoded by BC022504)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.37 mg/mL

          Amount

          55.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody got about 58kD in the mouse eye and 70kD in the mouse brain in western blotting, both in line with their expected masses. Defects in RGS9 are a cause of prolonged electroretinal response suppression (PERRS).

          Immunogen sequence: SGENLGFWE ACEDLKYGDQ SKVKEKAEEI YKLFLAPGAR RWINIDGKTM DITVKGLKHP HRYVLDAAQT HIYMLMKKDS YARYLKSPIY KDMLAKAIEP QETTKKSSTL PFMRRHLRSS PSPVILRQLE EEAKAREAAN TVDITQPGQH MAPSPHLTVY TGTCMPPSPS SPFSSSCRSP RKPFASPSRF IRRPSTTICP SPIRVALESS SGLEQKGECS GSMAPRGPSV TESSEASLDT SWPRSRPRAP PKARMALSFS RFLRRGCLAS PVFARLSPKC PAVSHGRVQP LGDVGQQLPR LKSKRVANFF QIKMDVPTGS GTCLMDSEDA GTGESGDRAT EKEVICPWES L (273-622 aa encoded by BC022504)

          Target Information

          The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alternatively spliced long form; MGC111763; MGC26458; Regulator of G-protein signaling 9; regulator of G-protein signalling 9; RGS9; RGS9L; ROS GAP; unnamed protein product

          View more View less

          Gene Aliases: PERRS; PERRS1; RGS9; RGS9-1; Rgs9-2; RGS9L

          View more View less

          UniProt ID: (Human) O75916, (Rat) P49805, (Mouse) O54828

          View more View less

          Entrez Gene ID: (Human) 8787, (Rat) 29481, (Mouse) 19739

          View more View less

          Function(s)
          GTPase activity GTPase activator activity protein binding signal transducer activity protein complex binding
          Process(es)
          response to amphetamine G-protein coupled receptor signaling pathway dopamine receptor signaling pathway nervous system development visual perception regulation of G-protein coupled receptor protein signaling pathway negative regulation of signal transduction response to estradiol intracellular signal transduction light adaption regulation of calcium ion export across plasma membrane dark adaptation termination of G-protein coupled receptor signaling pathway positive regulation of GTPase activity response to estrogen response to stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0