Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAIRKKLV VVGDGACGKT CLLIVFSKDE FPEVYVPTVF ENYVADIEVD GKQVELALWD TAGQEDYDRL RPLSYPDTDV ILMCFSVDSP DSLENIPEKW VPEVKHFCPN VPIILVANKK DLRSDEHVRT ELARMKQEPV RTDDGRAMAV RIQAYDYLEC SAKTKEGVRE VFETATRAAL QKRYGSQNGC INCCKVL (1-196 aa encoded by BC066954)
Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aplysia RAS-related homolog 6; aplysia ras-related homolog B; ARHB; h6; oncogene RHO H6; ras homolog B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; rho cDNA clone 6; rho protein; Rhob; small GTP binding protein; unnamed protein product
Gene Aliases: ARH6; ARHB; MST081; MSTP081; RHOH6
UniProt ID: (Human) P62745, (Rat) P62747, (Mouse) P62746
Entrez Gene ID: (Human) 388, (Rat) 64373, (Mouse) 11852
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support