Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RIM2 Antibodies

          Abnova

          RIMS2 Polyclonal Antibody, MaxPab™

          View all (11) RIM2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RIMS2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RIMS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RIMS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RIMS2 Antibody (H00009699-B01P) in WB

          Western Blot analysis of RIMS2 expression in transfected 293T cell line by RIMS2 MaxPab polyclonal antibody. Lane 1: RIMS2 transfected lysate(127.93 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RIMS2 Antibody in Western Blot (WB)

          Product Details

          H00009699-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RIMS2 (NP_055492.2, 1 a.a. approximately 1163 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQFETLRQVC NSVLSHFHGV FSSPPNILQN ELFGQTLNNA RKRSPSVSRD QNRRYDQREE REEYSQYATS DTAMPRSPSD YADRRFQHEP QFYEDSDHLS YRDSNRRSHR HSKEYIVDDE DVESRDEYER QRREEEYQSR YRSDPNLARY PVKPQPYEEQ MRIHAEVSRA RHERRHSDVS LANADLEDSR ISMLRMDRPS RQRSISERRA AMENQRSYSM ERTREAQGPS SYAQRTTNHS PPTPRRSPLP IDRPDLRRTD SLRKQHHLDP SSAVRKTKRE KMETMLRNDS LSSDRSESVR PPPPKPHKSK KGGKMRQISL SSSEEELAST PEYTSCDDVE IESESVSEKG DMDYNWLDHT SWHSSEASPM SLHPVTWQPS KDGDRLIGRI LLNKRLKDGS VPRDSGAMLG LKVVGGKMTE SGRLCAFITK VKKGSLADTV GHLRPGDEVL EWNGRLLQGA TFEEVYNIIL ESKPEPQVEL VVSRPIGDIP RIPDSTHAQL ESSSSSFESQ KMDRPSISVT SPMSPGMLRD VPQFLSGQLS IKLWFDKVGH QLIVTILGAK DLPSREDGRP RNPYVKIYFL PDRSDKNKRR TKTVKKTLEP KWNQTFIYSP VHRREFRERM LEITLWDQAR VREEESEFLG EILIELETAL LDDEPHWYKL QTHDVSSLPL PHPSPYMPRR QLHGESPTRR LQRSKRISDS EVSDYDCDDG IGVVSDYRHD GRDLQSSTLS VPEQVMSSNH CSPSGSPHRV DVIGRTRSWS PSVPPPQSRN VEQGLRGTRT MTGHYNTISR MDRHRVMDDH YSPDRDSHFL TLPRSRYSQT IDHHHRDGRD CEAADRQPYH RSRSTEQRPL LERTTTRSRS TERPDTNLMR SMPSLMTGRS APPSPALSRS HPRTGSVQTS PSSTPVAGRR GRQLPQLPPK GTLDRKAGGK KLRSTVQRST ETGLAVEMRN WMTRQASRES TDGSMNSYSS EGNLIFPGVR LASDSQFSDF LDGLGPAQLV GRQTLATPAM GDIQVGMMDK KGQLEVEIIR ARGLVVKPGS KTLPAPYVKV YLLDNGVCIA KKKTKVARKT LEPLYQQLLS FEESPQGKVL QIIVWGDYGR MDHKSFMGVA QILLDELELS NMVIGWFKLF PPSSLVDPTL APLTRRASQS SLESSTGPSY SRS

          Target Information

          Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: non-small cell lung cancer RIM2; non-small cell lung cancer RimL3a protein; non-small cell lung cancer RimL3c protein; nuclear protein; Rab-3-interacting molecule 2; Rab-3-interacting protein 3; RAB3 interacting protein 3; rab3-interacting molecule 2; Rab3-interacting protein; Regulating synaptic membrane exocytosis protein 2; RIM 2; RIM2 (CT); unnamed protein product

          View more View less

          Gene Aliases: CRSDS; KIAA0751; OBOE; RAB3IP3; RIM2; RIMS2

          View more View less

          UniProt ID: (Human) Q9UQ26

          View more View less

          Entrez Gene ID: (Human) 9699

          View more View less

          Function(s)
          protein binding zinc ion binding small GTPase binding ion channel binding metal ion binding structural constituent of presynaptic active zone
          Process(es)
          intracellular protein transport exocytosis adenylate cyclase-modulating G-protein coupled receptor signaling pathway synaptic vesicle docking synaptic vesicle priming calcium ion regulated exocytosis regulation of exocytosis insulin secretion cell differentiation regulation of membrane potential maintenance of presynaptic active zone structure calcium ion-regulated exocytosis of neurotransmitter spontaneous neurotransmitter secretion positive regulation of inhibitory postsynaptic potential positive regulation of dendrite extension regulation of synaptic vesicle exocytosis positive regulation of excitatory postsynaptic potential
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline