Search
Search
Fabgennix
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain-containing protein 3; PKC-associated protein kinase; PKC-delta-interacting protein kinase; PKC-regulated protein kinase; protein kinase C-associated kinase; serine/threonine-protein kinase ANKRD3; unnamed protein product
Gene Aliases: 2310069J12Rik; ANKK2; ANKRD3; CHANDS; DIK; NKRD3; PKK; PPS2; RIP4
UniProt ID: (Mouse) Q9ERK0
Entrez Gene ID: (Human) 54101, (Mouse) 72388, (Rat) 304053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support