Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RNF111 Antibodies

          Abnova

          RNF111 Polyclonal Antibody, MaxPab™

          View all (6) RNF111 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RNF111 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RNF111 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RNF111 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RNF111 Antibody (H00054778-B01P) in WB

          Western Blot analysis of RNF111 expression in transfected 293T cell line by RNF111 MaxPab polyclonal antibody. Lane 1: RNF111 transfected lysate(108.35 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RNF111 Antibody in Western Blot (WB)

          Product Details

          H00054778-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          3 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RNF111 (AAH60862.1, 1 a.a. approximately 985 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSQWTPEYKE LYTLKVDMKS EIPSDAPKTQ ESLKGILLHP EPIGAAKSFP AGVEMINSKV GNEFSHLCDD SQKQEKEMNG NQQEQEKSLV VRKKRKSQQA GPSYVQNCVK ENQGILGLRQ HLGTPSDEDN DSSFSDCLSS PSSSLHFGDS DTVTSDEDKE VSVRHSQTIL NAKSRSHSAR SHKWPRTETE SVSGLLMKRP CLHGSSLRRL PCRKRFVKNN SSQRTQKQKE RILMQRKKRE VLARRKYALL PSSSSSSEND LSSESSSSSS TEGEEDLFVS ASENHQNNPA VPSGSIDEDV VVIEASSTPQ VTANEEINVT STDSEVEIVT VGESYRSRST LGHSRSHWSQ GSSSHASRPQ EPRNRSRIST VIQPLRQNAA EVVDLTVDED EPTVVPTTSA RMESQATSAS INNSNPSTSE QASDTASAVT SSQPSTVSET SATLTSNSTT GTSIGDDSRR TTSSAVTETG PPAMPRLPSC CPQHSPCGGS SQNHHALGHP HTSCFQQHGH HFQHHHHHHH TPHPAVPVSP SFSDPACPVE RPPQVQAPCG ANSSSGTSYH EQALPVDLSN SGIRSHGSGS FHGASAFDPC CPVSSSRAAI FGHQAAAAAP SQPLSSIDGY GSSMVAQPQP QPPPQPSLSS CRHYMPPPYA SLTRPLHHQA SACPHSHGNP PPQTQPPPQV DYVIPHPVHA FHSQISSHAT SHPVAPPPPT HLASTAAPIP QHLPPTHQPI SHHIPATAPP AQRLHPHEVM QRMEVQRRRM MQHPTRAHER PPPHPHRMHP NYGHGHHIHV PQTMSSHPRQ APERSAWELG IEAGVTAATY TPGALHPHLA HYHAPPRLHH LQLGALPLMV PDMAGYPHIR YISSGLDGTS FRGPFRGNFE ELIHLEERLG NVNRGASQGT IERCTYPHKY KKRKLHCKQD GEEGTEEDTE EKCTICLSIL EEGEDVRRLP CMHLFHQVCV DQWLITNKKC PICRVDIEAQ LPSES

          Target Information

          The protein encoded by this gene is a nuclear RING-domain containing E3 ubiquitin ligase. This protein interacts with the transforming growth factor (TGF) -beta/NODAL signaling pathway by promoting the ubiquitination and proteosomal degradation of negative regulators, like SMAD proteins, and thereby enhances TGF-beta target-gene transcription. As a modulator of the nodal signaling cascade, this gene plays a critical role in the induction of mesoderm during embryonic development. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2012].

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Arkadia; E3 ubiquitin-protein ligase Arkadia; hRNF111; ring finger 111; RING finger protein 111; RING-type E3 ubiquitin transferase Arkadia; unnamed protein product

          View more View less

          Gene Aliases: ARK; hRNF111; RNF111

          View more View less

          UniProt ID: (Human) Q6ZNA4

          View more View less

          Entrez Gene ID: (Human) 54778, (Rat) 300813

          View more View less

          Function(s)
          protein binding zinc ion binding transferase activity SUMO polymer binding SMAD binding metal ion binding ubiquitin protein ligase activity ubiquitin-protein transferase activity
          Process(es)
          protein polyubiquitination pattern specification process protein ubiquitination positive regulation of transforming growth factor beta receptor signaling pathway ubiquitin-dependent SMAD protein catabolic process positive regulation of protein ubiquitination positive regulation of transcription, DNA-templated DNA repair ubiquitin-dependent protein catabolic process cellular response to DNA damage stimulus global genome nucleotide-excision repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline