PA1-31904 detects ZNF230 from human samples.
PA1-31904 has been successfully used in Western blot applications.
The PA1-31904 immunogen is: Synthetic peptide: LNMADEAGQPHRP from C Terminus of the protein sequence according to NP_057506.
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. Abundant expression of this gene was found in the testicular tissue of fertile men, but was not detected in azoospermic patients. Studies of the mouse counterpart suggest that this gene may function as a testis specific transcription factor during spermatogenesis.