Recommended positive controls: MCF-7.
Predicted reactivity: Rat (80%), Bovine (81%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 ubiquitin-protein ligase which promotes polyubiquitination and degradation by the proteasome pathway of ZIC2.