Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSLDIQSLD IQCEELSDAR WAELLPLLQQ CQVVRLDDCG LTEARCKDIS SALRVNPALA ELNLRSNELG DVGVHCVLQG LQTPSCKIQK LSLQNCCLTG AGCGVLSSTL RTLPTLQELH LSDNLLGDAG LQLLCEGLLD PQCRLEKLQL EYCSLSAASC EPLASVLRAK PDFKELTVSN NDINEAGVHV LCQGLKDSPC QLEALKLESC GVTSD (1-214 aa encoded by BC000677)
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: placental ribonuclease inhibitor; placental RNase inhibitor; ribonuclease inhibitor precursor; ribonuclease/angiogenin inhibitor; RNasIN; testicular tissue protein Li 164; unnamed protein product
Gene Aliases: IIAE12; RAI; RNH
UniProt ID: (Human) P13489, (Mouse) Q91VI7
Entrez Gene ID: (Human) 6050, (Mouse) 107702
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support