Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MARGPKKHL KRVAAPKHWM LDKLTGVFAP RPSTGPHKLR ECLPLIIFLR NRLKYALTGD EVKKICMQRF IKIDGKVRTD ITYPAGFMDV ISIDKTGENF RLIYDTKGRF AVHRITPEEA KYKLCKVRKI FVGTKGIPHL VTHDARTIRY PDPLIKVNDT IQIDLETGKI TDFIKFDTGN LCMVTGGANL GRIGVITNRE RHPGSFDVVH VKDANGNSFA TRLSNIFVIG KGNKPWISLP RGKGIRLTIA EERDKRLAAK QSSG (1-263 aa encoded by BC000472)
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S4; cell cycle gene 2; ribosomal protein S4, X-linked X-like; RPS4X; scar protein; single copy abundant mRNA protein; single-copy abundant mRNA; small ribosomal subunit protein eS4; unnamed protein product
Gene Aliases: CCG2; DXS306; eS4; RPS4; Rps4-1; S4; SCAR; SCR10
UniProt ID: (Human) P62701, (Mouse) P62702, (Rat) P62703
Entrez Gene ID: (Human) 6191, (Mouse) 20102, (Rat) 100362640
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support