Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: IRNSLLIAP MPTASTAQIL GNNESIEPYT SNIYTRRVLS GEFQIVNPHL LKDLTERGLW HEEMKNQIIA CNGSIQSIPE IPDDLKQLYK TVWEISQKTV LKMAAERGAF IDQSQSLNIH IAEPNYGKLT SMHFYGWKQG LKTGMYYLRT RPAANPIQFT LNKEKLKDKE KVSKEEEEKE RNTAAMVCSL ENRDECLMCG S (593-792 aa encoded by BC006498)
RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: R RRM 1; RI RI RR 1; ribonucleoside-diphosphate reductase subunit M1; ribonucleotide reductase large subunit; ribonucleotide reductase M1; ribonucleotide reductase M1 polypeptide; ribonucleotide reductase subunit M1; ribonucleotide reductase, R1 subunit; RIR 1; unnamed protein product
Gene Aliases: PEOB6; R1; RIR1; RnrM1; RR1
UniProt ID: (Human) P23921, (Mouse) P07742
Entrez Gene ID: (Human) 6240, (Mouse) 20133, (Rat) 685579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support