Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 11
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Predicted reactivity: Mouse (89%), Rat (88%), Zebrafish (86%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: etID309896.20; reductase M2 polypeptide variant 1; reductase M2 polypeptide variant 2; reductase M2 polypeptide variant 3a; reductase M2 polypeptide variant 3b; reductase M2 polypeptide variant 3c; reductase M2 polypeptide variant 3d; ribonucleoside-diphosphate reductase M2 chain; Ribonucleotide reductase 2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase protein r2 class I; ribonucleotide reductase small chain; ribonucleotide reductase small subunit; ribonucleotide reductase subunit M2; RRR2M; uncharacterized protein C2orf48; unnamed protein product
Gene Aliases: C2orf48; cb111; chunp6884; R2; RR2; RR2M
UniProt ID: (Human) P31350, (Rat) Q4KLN6, (Mouse) P11157, (Zebrafish) P79733
Entrez Gene ID: (Human) 6241, (Rat) 362720, (Mouse) 20135, (Zebrafish) 30733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support