Immunogen sequence: DQIMMCSMYG ICKVKNIDLK FKIIVTAYKD LPHAVQETFK RVLIKEEEYD SIIVFYNSVF MQRLKTNILQ YASTRPPTLS PIPHIPRSPY KFPSSPLRIP GGNIYISPLK SPYKISEGLP TPTKMTPRSR ILVSIGESFG TSEKFQKINQ MVCNSDRVLK RSAEGSNPPK PLKKLRFDIE GSDEADGSKH LPGESKFQQK LAEMTSTRTR MQKQKMNDSM DTSNKEEK
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.