Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Reelin Antibodies

          Invitrogen

          Reelin Polyclonal Antibody

          View all (13) Reelin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Reelin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Reelin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Reelin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Reelin Antibody (PA5-104090) in WB

          Western blot analysis of Reelin in RAW264.7 cell lysate. Samples were incubated with Reelin polyclonal antibody (Product # PA5-104090). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Reelin Antibody in Western Blot (WB)
          Reelin Polyclonal Antibody

          Product Details

          PA5-104090

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human RELN(Accession P78509), corresponding to amino acid residues W294-A344.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2853418

          Product Specific Information

          Antibody detects endogenous levels of total RELN.

          Target Information

          Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: extracellular matrix serine protease; Reeler protein; Reelin; unnamed protein product

          View more View less

          Gene Aliases: ETL7; LIS2; PRO1598; Reelen; reeler; RELN; RL

          View more View less

          UniProt ID: (Human) P78509, (Mouse) Q60841, (Rat) P58751

          View more View less

          Entrez Gene ID: (Human) 5649, (Mouse) 19699, (Rat) 24718

          View more View less

          Function(s)
          peptidase activity serine-type peptidase activity hydrolase activity metal ion binding receptor agonist activity lipoprotein particle receptor binding very-low-density lipoprotein particle receptor binding protein binding molecular_function protein serine/threonine/tyrosine kinase activity cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein
          Process(es)
          cell morphogenesis neuron migration proteolysis cell adhesion axon guidance central nervous system development brain development learning long-term memory locomotory behavior associative learning glial cell differentiation regulation of gene expression positive regulation of neuron projection development dendrite development cell migration spinal cord patterning ventral spinal cord development hippocampus development cerebral cortex tangential migration layer formation in cerebral cortex cerebral cortex development forebrain development positive regulation of TOR signaling protein localization to synapse reelin-mediated signaling pathway proteasome-mediated ubiquitin-dependent protein catabolic process regulation of neuron differentiation response to pain regulation of behavior modulation of synaptic transmission positive regulation of small GTPase mediated signal transduction positive regulation of protein kinase B signaling positive regulation of synaptic transmission, glutamatergic regulation of synaptic activity long-term synaptic potentiation positive regulation of dendritic spine morphogenesis regulation of synapse maturation positive regulation of synapse maturation postsynaptic density assembly NMDA glutamate receptor clustering postsynaptic density protein 95 clustering receptor localization to synapse motor neuron migration lateral motor column neuron migration positive regulation of long-term synaptic potentiation positive regulation of lateral motor column neuron migration interneuron migration positive regulation of excitatory postsynaptic potential regulation of neuron migration cell morphogenesis involved in differentiation multicellular organism development positive regulation of phosphatidylinositol 3-kinase signaling peptidyl-tyrosine phosphorylation dentate gyrus development positive regulation of CREB transcription factor activity thyroid hormone metabolic process positive regulation of protein kinase activity positive regulation of peptidyl-tyrosine phosphorylation positive regulation of protein tyrosine kinase activity regulation of N-methyl-D-aspartate selective glutamate receptor activity positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity midgut development memory response to mechanical stimulus response to toxic substance response to hormone response to lithium ion response to organic cyclic compound response to insecticide orbitofrontal cortex development hypothalamus development response to progesterone social behavior response to drug response to amino acid response to ethanol response to axon injury response to corticosterone response to electrical stimulus response to fatty acid cellular response to dexamethasone stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline