Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • RhoA Antibodies

          Invitrogen

          RhoA Polyclonal Antibody

          View all (28) RhoA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RhoA Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          RhoA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RhoA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RhoA Antibody (PA5-120243) in IHC (P)

          Immunohistochemistry analysis of RhoA in paraffin-embedded human placenta. Samples were incubated with RhoA Polyclonal antibody (Product # PA5-120243) using a dilution of 1:100 (40x lens). Perform microwave antigen retrieval with 10 mM Tris/EDTA buffer pH 9.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RhoA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA Antibody in Western Blot (WB)
          RhoA Polyclonal Antibody

          Product Details

          PA5-120243

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 94-193 of human RhoA (NP_001655.1).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2.77 mg/mL

          Amount

          277 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2913815

          Product Specific Information

          Positive test controls include: NIH/3T3, Mouse lung. The target is usually found in the following locations: Cell membrane, Cell projection, Cleavage furrow, Cytoplasm, Cytoplasmic side, Lipid-anchor, Midbody, cell cortex, cytoskeleton, lamellipodium.

          Immunogen sequence: NIPEKWTPEV KHFCPNVPII LVGNKKDLRN DEHTRRELAK MKQEPVKPEE GRDMANRIGA FGYMECSAKT KDGVREVFEM ATRAALQARR GKKKSGCLVL

          Target Information

          RhoA regulates a signal transduction pathway linking plasma membrane receptors to the assembly of focal adhesions and actin stress fibers. It is involved in a microtubule-dependent signal that is required for the myosin contractile ring formation during cell cycle cytokinesis, and plays an essential role in cleavage furrow formation. RhoA is required for the apical junction formation of keratinocyte cell-cell adhesion, and serves as a target for the yopT cysteine peptidase from Yersinia pestis, vector of the plague, and Yersinia pseudotuberculosis, which causes gastrointestinal disorders. RhoA stimulates PKN2 kinase activity and may be an activator of PLCE1. It is activated by ARHGEF2, which promotes the exchange of GDP for GTP. It is essential for the SPATA13-mediated regulation of cell migration and adhesion assembly and disassembly. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Aplysia ras-related homolog 12; aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; epididymis secretory sperm binding protein; h12; MGC72339; oncogene RHO H12; plysia ras-related homolog A2; Ras family member A; ras homolog A1; ras homolog A2; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; Rho; Rho cDNA clone 12; Rho family GTPase; small GTP binding protein RhoA; Transforming protein RhoA; unnamed protein product

          View more View less

          Gene Aliases: ARH12; ARHA; Arha1; Arha2; EDFAOB; RHO12; RHOA; RHOH12

          View more View less

          UniProt ID: (Human) P61586, (Rat) P61589, (Mouse) Q9QUI0

          View more View less

          Entrez Gene ID: (Human) 387, (Rat) 117273, (Mouse) 11848

          View more View less

          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding hydrolase activity myosin binding protein kinase binding GDP binding protein domain specific binding Rho GDP-dissociation inhibitor binding small GTPase
          Process(es)
          mitotic cytokinesis cell morphogenesis neuron migration kidney development angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure alpha-beta T cell lineage commitment regulation of systemic arterial blood pressure by endothelin aortic valve formation regulation of transcription from RNA polymerase II promoter cytoskeleton organization cell adhesion cell-matrix adhesion small GTPase mediated signal transduction Rho protein signal transduction skeletal muscle tissue development negative regulation of cell-substrate adhesion regulation of neuron projection development cell migration substantia nigra development cerebral cortex cell migration forebrain radial glial cell differentiation actin cytoskeleton organization cell differentiation regulation of cell migration positive regulation of cell migration androgen receptor signaling pathway cytoplasmic microtubule organization positive regulation of cytokinesis regulation of actin cytoskeleton organization negative regulation of intracellular steroid hormone receptor signaling pathway regulation of osteoblast proliferation cell junction assembly substrate adhesion-dependent cell spreading Roundabout signaling pathway cleavage furrow formation apolipoprotein A-I-mediated signaling pathway odontogenesis positive regulation of I-kappaB kinase/NF-kappaB signaling stress fiber assembly apical junction assembly beta selection endothelial cell migration ossification involved in bone maturation wound healing, spreading of cells establishment of epithelial cell apical/basal polarity positive regulation of neuron differentiation negative regulation of cell size positive regulation of alpha-beta T cell differentiation negative chemotaxis cell division positive regulation of stress fiber assembly regulation of focal adhesion assembly Wnt signaling pathway, planar cell polarity pathway positive regulation of lipase activity bone trabecula morphogenesis regulation of microtubule cytoskeleton organization cellular response to lipopolysaccharide cellular response to cytokine stimulus semaphorin-plexin signaling pathway positive regulation of podosome assembly positive regulation of protein serine/threonine kinase activity negative regulation of cell migration involved in sprouting angiogenesis mitotic spindle assembly negative regulation of oxidative phosphorylation motor neuron apoptotic process endothelial tube lumen extension regulation of modification of postsynaptic structure positive regulation of NIK/NF-kappaB signaling skeletal muscle satellite cell migration negative regulation of reactive oxygen species biosynthetic process mitotic cleavage furrow formation positive regulation of vascular smooth muscle contraction positive regulation of leukocyte adhesion to vascular endothelial cell regulation of modification of postsynaptic actin cytoskeleton cellular response to chemokine regulation of neural precursor cell proliferation positive regulation of T cell migration negative regulation of motor neuron apoptotic process response to hypoxia regulation of actin polymerization or depolymerization metabolic process response to mechanical stimulus response to glucose positive regulation of cell growth positive regulation of actin filament polymerization stress-activated protein kinase signaling cascade response to drug negative regulation of I-kappaB kinase/NF-kappaB signaling response to amino acid positive regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process response to ethanol negative regulation of neuron differentiation positive regulation of translation positive regulation of cell adhesion positive regulation of vasoconstriction positive regulation of smooth muscle contraction neuron projection morphogenesis regulation of dendrite development response to glucocorticoid regulation of calcium ion transport negative regulation of cell death trabecula morphogenesis cellular response to progesterone stimulus cell cycle integrin-mediated signaling pathway GTP metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0