Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • RhoA/RhoB/RhoC Antibodies

          Invitrogen

          RhoA/RhoB/RhoC Recombinant Rabbit Monoclonal Antibody (5E6)

          View all (5) RhoA/RhoB/RhoC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RhoA/RhoB/RhoC Recombinant Rabbit Monoclonal Antibody (5E6)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (16)
          RhoA/RhoB/RhoC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RhoA/RhoB/RhoC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RhoA/RhoB/RhoC Antibody (MA572483) in ICC/IF

          Immunocytochemistry analysis of RhoA/RhoB/RhoC in HepG2 cells. Permeabilized with Trinton X-100 at room temp for 20 min. Samples were incubated with RhoA/RhoB/RhoC Recombinant Monoclonal (Product # MA572483 using a dilution at 1:100, for 90 min at 37°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (green) at 37°C for 90 min, DAPI was used to stain the cell nuclei. PBS instead of the primary antibody... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RhoA/RhoB/RhoC Antibody in Immunocytochemistry (ICC/IF)
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RhoA/RhoB/RhoC Antibody in Western Blot (WB)
          RhoA/RhoB/RhoC Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA572483

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5E6

          Immunogen

          KLH conjugated synthetic peptide derived from human RhoA.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog B; ARHB; ARHC; epididymis secretory sperm binding protein; h6; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; ras homolog 9 (RhoC); ras homolog B; ras homolog gene family, member A; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; RAS-related homolog 9; Rho; rho cDNA clone 6; rho cDNA clone 9; rho protein; RhoA; Rhob; rhoC GTPase; SIG-61; silica-induced gene 61 protein; small GTP binding protein; small GTP binding protein RhoA; small GTP binding protein RhoC; unnamed protein product

          View more View less

          Gene Aliases: ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; ARHB; ARHC; EDFAOB; H9; MST081; MSTP081; RHO12; RHOH12; RHOH6; RHOH9

          View more View less

          UniProt ID: (Human) P61586, (Human) P62745, (Human) P08134, (Rat) P61589, (Mouse) Q9QUI0, (Rat) P62747, (Mouse) P62746, (Mouse) Q62159

          View more View less

          Entrez Gene ID: (Human) 387, (Human) 388, (Human) 389, (Rat) 117273, (Mouse) 11848, (Rat) 64373, (Mouse) 11852, (Rat) 295342, (Mouse) 11853

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding myosin binding protein kinase binding GDP binding protein domain specific binding Rho GDP-dissociation inhibitor binding
          Process(es)
          mitotic cytokinesis actin filament organization cell adhesion signal transduction small GTPase mediated signal transduction endosome to lysosome transport positive regulation of endothelial cell migration regulation of cell migration negative regulation of cell migration positive regulation of apoptotic process positive regulation of angiogenesis negative regulation of cell cycle endothelial tube morphogenesis cellular response to hydrogen peroxide cellular response to ionizing radiation regulation of modification of postsynaptic structure negative regulation of establishment of endothelial barrier alpha-beta T cell lineage commitment aortic valve formation Rho protein signal transduction negative regulation of cell-substrate adhesion cell migration substantia nigra development actin cytoskeleton organization positive regulation of cell migration cytoplasmic microtubule organization positive regulation of cytokinesis regulation of actin cytoskeleton organization regulation of osteoblast proliferation cell junction assembly substrate adhesion-dependent cell spreading Roundabout signaling pathway cleavage furrow formation apolipoprotein A-I-mediated signaling pathway odontogenesis positive regulation of I-kappaB kinase/NF-kappaB signaling stress fiber assembly apical junction assembly beta selection endothelial cell migration ossification involved in bone maturation wound healing, spreading of cells establishment of epithelial cell apical/basal polarity positive regulation of neuron differentiation negative regulation of cell size positive regulation of alpha-beta T cell differentiation negative chemotaxis positive regulation of stress fiber assembly regulation of focal adhesion assembly Wnt signaling pathway, planar cell polarity pathway positive regulation of lipase activity bone trabecula morphogenesis regulation of microtubule cytoskeleton organization cellular response to lipopolysaccharide cellular response to cytokine stimulus semaphorin-plexin signaling pathway positive regulation of protein serine/threonine kinase activity negative regulation of cell migration involved in sprouting angiogenesis mitotic spindle assembly negative regulation of oxidative phosphorylation endothelial tube lumen extension positive regulation of NIK/NF-kappaB signaling skeletal muscle satellite cell migration negative regulation of reactive oxygen species biosynthetic process mitotic cleavage furrow formation positive regulation of leukocyte adhesion to vascular endothelial cell regulation of modification of postsynaptic actin cytoskeleton cellular response to chemokine positive regulation of T cell migration intracellular protein transport positive regulation of protein complex assembly meiotic spindle organization cell morphogenesis response to hypoxia kidney development angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure regulation of systemic arterial blood pressure by endothelin regulation of transcription from RNA polymerase II promoter cytoskeleton organization microtubule depolymerization cell-matrix adhesion skeletal muscle tissue development regulation of actin polymerization or depolymerization response to xenobiotic stimulus response to mechanical stimulus response to glucose regulation of neuron projection development negative regulation of neuron projection development cerebral cortex cell migration forebrain radial glial cell differentiation cell differentiation positive regulation of cell growth androgen receptor signaling pathway positive regulation of actin filament polymerization establishment or maintenance of actin cytoskeleton polarity stress-activated protein kinase signaling cascade negative regulation of intracellular steroid hormone receptor signaling pathway embryonic cleavage polar body extrusion after meiotic divisions negative regulation of I-kappaB kinase/NF-kappaB signaling response to amino acid negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process response to ethanol negative regulation of neuron differentiation positive regulation of translation positive regulation of cell adhesion positive regulation of vasoconstriction positive regulation of smooth muscle contraction GTP metabolic process neuron projection morphogenesis regulation of dendrite development response to glucocorticoid spindle localization regulation of calcium ion transport negative regulation of synapse assembly cellular response to progesterone stimulus positive regulation of podosome assembly positive regulation of vascular smooth muscle contraction negative regulation of vascular smooth muscle cell proliferation negative regulation of vascular associated smooth muscle cell migration regulation of neural precursor cell proliferation apoptotic process integrin-mediated signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-dlj7x:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0