Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calgranulin A; calgranulin-A; calprotectin L1L subunit; chemotactic cytokine CP-10; chemotactic S100 protein; cystic fibrosis antigen; leukocyte L1 complex light chain; migration inhibitory factor-related protein 8; MRP-8; pro-inflammatory S100 cytokine; S100 A8; S100 calcium-binding protein A8; S100A8; unnamed protein product; urinary stone protein band A
Gene Aliases: 60B8AG; B8Ag; CAGA; CFAG; CGLA; CP-10; L1Ag; MA387; MRP8; NIF; P8; S100-A8
UniProt ID: (Human) P05109, (Mouse) P27005
Entrez Gene ID: (Human) 6279, (Mouse) 20201
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support