    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
            • Custom Products & Projects
          • Secondary Antibodies ›
          • Rabbit anti-Human IgE Secondary Antibodies
          Invitrogen
          Rabbit anti-Human IgE Recombinant Secondary Antibody
          Advanced Verification
          View all (2) Rabbit anti-Human IgE antibodies
          Tech Support
          Cite Rabbit anti-Human IgE Recombinant Secondary Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          • Advanced Verification (1)
          Human IgE Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Human IgE Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          Human IgE Secondary Antibody (SA5-10201) in WB
          Western blot was performed using Rabbit anti-Human IgE Recombinant Secondary Antibody (Product # SA5-10201) and a 73 kDa band corresponding to Human IgE Heavy Chain was observed in IgE but not in IgG, IgA, IgM, Mouse IgG, IgM and Rat IgG. Purified protein (100 ng) of Human IgE (Lane 1), IgG (Lane 2), IgA (Lane 3), IgM (Lane 4), Mouse IgG (Lane 5), IgM (Lane 6) and Rat IgG (Lane 7) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human IgE Secondary Antibody in Western Blot (WB)
          Human IgE Secondary Antibody in ELISA (ELISA)
          Human IgE Secondary Antibody in ELISA (ELISA)
          Human IgE Secondary Antibody in ELISA (ELISA)
          Human IgE Secondary Antibody
          Product Details
          SA5-10201
          Applications
          Tested Dilution
          Western Blot (WB)
          1:5,000
          ELISA (ELISA)
          10-100 ng/well (Capture), 0.01-0.1 µg/mL (Detect)
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Secondary Antibody
          Clone
          RM122
          Immunogen
          Peptide corresponding to Human IgE.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2-7.4, with 1% BSA, 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2665330
          Target
          IgE
          Antibody Form
          Whole Antibody
          Product Specific Information
          This antibody reacts to human IgE. No cross reactivity with human IgG, IgM, IgD, or IgA.
          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
          Target Information
          Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody SA5-10201 on CiteAb
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline