    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Rat IgG2a Secondary Antibodies
          Invitrogen
          Goat anti-Rat IgG2a Recombinant Secondary Antibody, Alexa Fluor™ Plus 555
          Advanced Verification
          View all (8) Goat anti-Rat IgG2a antibodies
          Tech Support
          Cite Goat anti-Rat IgG2a Recombinant Secondary Antibody, Alexa Fluor™ Plus 555
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          Rat IgG2a Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rat IgG2a Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Rat IgG2a Secondary Antibody (SA5-10376-AFP555) in ICC/IF
          Immunofluorescence analysis of Goat anti-Rat IgG2a Recombinant Secondary Antibody, Alexa Fluor™ Plus 555 (Product # SA5-10376-AFP555) was performed on fixed and permeabilized LNCaP cells blocked with SuperBlock™ Blocking Buffer at room temperature and stained with alpha Tubulin Monoclonal Antibody (YL1/2) (Product # MA1-80017, 1:500) in SuperBlock™ Blocking Buffer (Product # 37515, 1:20) overnight at 4 degr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Rat IgG2a Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rat IgG2a Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rat IgG2a Secondary Antibody
          Product Details
          SA5-10376-AFP555
          Applications
          Tested Dilution
          Immunohistochemistry (Paraffin) (IHC (P))
          1:400
          Immunocytochemistry (ICC/IF)
          1:1,000
          Product Specifications
          Species Reactivity
          Rat
          Host/Isotype
          Goat / IgG
          Expression System
          Expi293
          Class
          Recombinant Monoclonal
          Type
          Secondary Antibody
          Clone
          3H24L4
          Immunogen
          Purified Rat IgG
          Conjugate
          Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555 Alexa Fluor™ Plus 555
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 680 
          • FITC
          • HRP
          • PE 
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          558/572 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          250 µg
          Purification
          Affinity chromatography
          Storage buffer
          proprietary buffer, pH 6.3-7.3
          Contains
          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          Target
          IgG2a
          Antibody Form
          Whole Antibody
          Product Specific Information
          Goat anti-Rat IgG2a Recombinant Secondary Antibody, Alexa Fluor™ Plus 555 is specific to Rat IgG2a. It has ≤ 5% cross reactivity to Mouse IgG and Rabbit IgG. It showed no cross-reactivity to various species immunoglobulins tested.
          Alexa Fluor™ Plus dyes have been chemically optimized to provide higher signal to noise ratios and minimized cross-reactivity to help detect low-abundance targets in rare or precious samples.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Target Information
          Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody SA5-10376-AFP555 on CiteAb
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0