    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Rat IgG2b Secondary Antibodies
          Invitrogen
          Goat anti-Rat IgG2b Recombinant Secondary Antibody, Alexa Fluor™ Plus 680
          Advanced Verification
          View all (6) Goat anti-Rat IgG2b antibodies
          Tech Support
          Cite Goat anti-Rat IgG2b Recombinant Secondary Antibody, Alexa Fluor™ Plus 680
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          Rat IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rat IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Rat IgG2b Secondary Antibody (SA5-10377-AFP680) in ICC/IF
          Immunofluorescence analysis of Goat anti-Rat IgG2b, Recombinant Secondary Antibody, Alexa Fluor™ Plus 680 (Product # SA5-10377-AFP680) was performed on fixed and permeabilized HeLa cells. Cells were blocked with SuperBlock™ Blocking Buffer for 1 hour at room temperature and probed with hnRNP UL1 Monoclonal Antibody (EAP 4A11) (Product # 14-9806-82, 1:400) overnight at 4 degrees Celsius. This was detected by... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Rat IgG2b Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rat IgG2b Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Rat IgG2b Secondary Antibody
          Product Details
          SA5-10377-AFP680
          Applications
          Tested Dilution
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500
          Immunocytochemistry (ICC/IF)
          1:1,000
          Product Specifications
          Species Reactivity
          Rat
          Host/Isotype
          Goat / IgG
          Expression System
          Expi293
          Class
          Recombinant Monoclonal
          Type
          Secondary Antibody
          Clone
          10H38L66
          Immunogen
          Purified Rat IgG
          Conjugate
          Alexa Fluor™ Plus 680 Alexa Fluor™ Plus 680 Alexa Fluor™ Plus 680
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • HRP
          • PE 
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          687/704 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          250 µg
          Purification
          Affinity chromatography
          Storage buffer
          proprietary buffer, pH 6.3-7.3
          Contains
          0.008% Bromonitrodioxane, 0.008% Methylisothiazolone
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          RRID
          AB_3716377
          Target
          IgG2b
          Antibody Form
          Whole Antibody
          Product Specific Information
          Goat anti-Rat IgG2b, Recombinant Secondary Antibody specifically detects the Fc portion of rat IgG2b isotype and shows strong reactivity to total rat IgG. It has minimal to no cross-reactivity towards other rat IgG isotypes and immunoglobulins tested from other species.
          Alexa Fluor™ Plus conjugated recombinant antibodies provide bright signals and high signal-to-noise for your critical experiments.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Target Information
          Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody SA5-10377-AFP680 on CiteAb
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-fcf5f9d6d-kppxg:80/100.66.77.222:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0