Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • SAP97 Antibodies

          Bioss

          SAP97 Polyclonal Antibody

          View all (21) SAP97 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SAP97 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          SAP97 Polyclonal Antibody

          Product Details

          BS-11361R

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human SAP97, amino acids 801-904.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: discs large homolog 1, scribble cell polarity complex component; Disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; presynaptic protein SAP97; SAP-97; Synapse-associated protein 97; unnamed protein product

          View more View less

          Gene Aliases: B130052P05Rik; DLG1; DLGH1; E-dlg/SAP97; hdlg; mKIAA4187; SAP-97; SAP97

          View more View less

          UniProt ID: (Human) Q12959, (Rat) Q62696, (Mouse) Q811D0

          View more View less

          Entrez Gene ID: (Human) 1739, (Rat) 25252, (Mouse) 13383

          View more View less

          Function(s)
          guanylate kinase activity phosphoprotein phosphatase activity protein binding cytoskeletal protein binding potassium channel regulator activity kinase binding protein kinase binding phosphatase binding ionotropic glutamate receptor binding ion channel binding cadherin binding binding, bridging L27 domain binding structural constituent of postsynaptic density protein C-terminus binding PDZ domain binding mitogen-activated protein kinase kinase binding protein complex scaffold scaffold/adaptor protein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ureteric bud development branching involved in ureteric bud morphogenesis immunological synapse formation endothelial cell proliferation lens development in camera-type eye actin filament organization establishment or maintenance of cell polarity chemical synaptic transmission nervous system development protein localization cell proliferation positive regulation of cell proliferation regulation of cell shape actin filament polymerization peristalsis positive regulation of actin filament polymerization cortical actin cytoskeleton organization astral microtubule organization protein complex localization membrane raft organization regulation of myelination regulation of protein localization protein localization to synapse T cell proliferation T cell activation negative regulation of T cell proliferation regulation of membrane potential amyloid precursor protein metabolic process receptor clustering regulation of potassium ion transport positive regulation of potassium ion transport protein kinase B signaling cortical microtubule organization establishment or maintenance of epithelial cell apical/basal polarity GMP metabolic process GDP metabolic process reproductive structure development embryonic skeletal system morphogenesis tissue morphogenesis smooth muscle tissue development negative regulation of epithelial cell proliferation establishment of centrosome localization negative regulation of protein kinase B signaling hard palate development negative regulation of ERK1 and ERK2 cascade bicellular tight junction assembly protein localization to membrane protein localization to plasma membrane receptor localization to synapse cell-cell adhesion regulation of ventricular cardiac muscle cell action potential membrane repolarization during ventricular cardiac muscle cell action potential regulation of postsynaptic membrane neurotransmitter receptor levels maintenance of postsynaptic density structure neurotransmitter receptor localization to postsynaptic specialization membrane regulation of NIK/NF-kappaB signaling regulation of sodium ion transmembrane transport regulation of protein localization to synapse positive regulation of protein localization to plasma membrane regulation of potassium ion import negative regulation of p38MAPK cascade regulation of potassium ion export across plasma membrane protein localization to cell periphery negative regulation of G1/S transition of mitotic cell cycle T cell cytokine production cell adhesion single organismal cell-cell adhesion activation of protein kinase activity negative regulation of mitotic cell cycle nucleotide phosphorylation positive regulation of establishment of protein localization to plasma membrane positive regulation of multicellular organism growth positive regulation of developmental growth
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0