Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SCN2B Antibodies

          Alomone Labs, LTD

          SCN2B (NaV beta 2) Polyclonal Antibody

          View all (13) SCN2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SCN2B (NaV beta 2) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          SCN2B (NaV beta 2) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          SCN2B (NaV beta 2) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SCN2B (NaV beta 2) Antibody (ASC-007-200UL) in IHC

          Expression of NaV beta 2in mouse cerebellum - Immunohistochemical staining of mouse cerebellum using Anti-SCN2B (NaV beta 2) Antibody (#ASC-007). A. NaV beta 2channel (red) appears in Purkinje cells (vertical arrows) and in small cells inthe molecular layer (horizontal arrows, Mol). B. Staining of Purkinje nerve cells with mouse Anti-calcium binding protein (CBD28k, green) demonstrates the restriction of Na... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SCN2B (NaV beta 2) Antibody in Immunohistochemistry (IHC)
          SCN2B (NaV beta 2) Antibody in Immunohistochemistry (IHC)
          SCN2B (NaV beta 2) Antibody in Western Blot (WB)
          SCN2B (NaV beta 2) Polyclonal Antibody

          Product Details

          ASC-007-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)KTEEEGKTDGEGNAEDGAK, corresponding to amino acid residues 197-215 of rat beta 2subunit of voltage-gated Na+channels, Intracellular, C-terminus
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.75 mg/mL

          Amount

          150 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Scn2b is a member of voltage-gated sodium channels (NaV) that are responsible for action potential initiation and propagation in excitable cells, including nerve, muscle, and neuroendocrine cell types. Scn2b are also expressed at low levels in non-excitable cells, where their physiological role is unclear. Scn2b is crucial in the assembly, expression, and functional modulation of the heterotrimeric complex of the sodium channel. In Scn2b, the subunit beta-2 causes an increase in the plasma membrane surface area and in its folding into microvilli. Also, Scn2b interacts with TNR (Tenascin R-precursor) and may play a crucial role in clustering and regulation of activity of sodium channels at nodes of Ranvier. The protein encoded by the Scn2b gene is the beta 2 subunit of the type II voltage-gated sodium channel. Scn2b protein is involved in cell-cell adhesion and cell migration. Defects in the Scn2b gene can be a cause of Brugada Syndrome, atrial fibrillation, or sudden infant death syndrome. Diseases associated with SCN2B include Atrial Fibrillation, Familial, 14 and Scn2b-Related Familial Atrial Fibrillation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: neuronal voltage-gated sodium channel beta 2 subunit; Sodium channel beta 2; Sodium channel regulatory subunit beta-2; sodium channel subunit beta-2; sodium channel, voltage gated, type II beta subunit; sodium channel, voltage-gated, type II, beta polypeptide; unnamed protein product

          View more View less

          Gene Aliases: 2810451E09Rik; AI840361; ATFB14; Gm183; SCN2B; SCNB2; UNQ326/PRO386

          View more View less

          UniProt ID: (Human) O60939, (Rat) P54900, (Mouse) Q56A07

          View more View less

          Entrez Gene ID: (Human) 6327, (Rat) 25349, (Mouse) 72821

          View more View less

          Function(s)
          voltage-gated sodium channel activity sodium channel regulator activity voltage-gated sodium channel activity involved in cardiac muscle cell action potential voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization protein binding voltage-gated ion channel activity sodium channel activity myelin protein
          Process(es)
          ion transport sodium ion transport chemical synaptic transmission nervous system development response to heat gene expression positive regulation of sodium ion transport sodium ion transmembrane transport response to pyrethroid cardiac muscle contraction regulation of atrial cardiac muscle cell membrane depolarization cardiac conduction cardiac muscle cell action potential involved in contraction membrane depolarization during action potential membrane depolarization during cardiac muscle cell action potential regulation of heart rate by cardiac conduction membrane repolarization during ventricular cardiac muscle cell action potential regulation of sodium ion transmembrane transporter activity transport regulation of ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline