Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SCYL2 Antibodies

          Abnova

          SCYL2 Polyclonal Antibody, MaxPab™

          View all (6) SCYL2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SCYL2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SCYL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SCYL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SCYL2 Antibody (H00055681-D01P) in WB

          SCYL2 MaxPab rabbit polyclonal antibody. Western Blot analysis of SCYL2 expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SCYL2 Antibody in Western Blot (WB)
          SCYL2 Polyclonal Antibody, MaxPab™

          Product Details

          H00055681-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SCYL2 (NP_060458.3, 1 a.a. approximately 929 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MESMLNKLKS TVTKVTADVT SAVMGNPVTR EFDVGRHIAS GGNGLAWKIF NGTKKSTKQE VAVFVFDKKL IDKYQKFEKD QIIDSLKRGV QQLTRLRHPR LLTVQHPLEE SRDCLAFCTE PVFASLANVL GNWENLPSPI SPDIKDYKLY DVETKYGLLQ VSEGLSFLHS SVKMVHGNIT PENIILNKSG AWKIMGFDFC VSSTNPSEQE PKFPCKEWDP NLPSLCLPNP EYLAPEYILS VSCETASDMY SLGTVMYAVF NKGKPIFEVN KQDIYKSFSR QLDQLSRLGS SSLTNIPEEV REHVKLLLNV TPTVRPDADQ MTKIPFFDDV GAVTLQYFDT LFQRDNLQKS QFFKGLPKVL PKLPKRVIVQ RILPCLTSEF VNPDMVPFVL PNVLLIAEEC TKEEYVKLIL PELGPVFKQQ EPIQILLIFL QKMDLLLTKT PPDEIKNSVL PMVYRALEAP SIQIQELCLN IIPTFANLID YPSMKNALIP RIKNACLQTS SLAVRVNSLV CLGKILEYLD KWFVLDDILP FLQQIPSKEP AVLMGILGIY KCTFTHKKLG ITKEQLAGKV LPHLIPLSIE NNLNLNQFNS FISVIKEMLN RLESEHKTKL EQLHIMQEQQ KSLDIGNQMN VSEEMKVTNI GNQQIDKVFN NIGADLLTGS ESENKEDGLQ NKHKRASLTL EEKQKLAKEQ EQAQKLKSQQ PLKPQVHTPV ATVKQTKDLT DTLMDNMSSL TSLSVSTPKS SASSTFTSVP SMGIGMMFST PTDNTKRNLT NGLNANMGFQ TSGFNMPVNT NQNFYSSPST VGVTKMTLGT PPTLPNFNAL SVPPAGAKQT QQRPTDMSAL NNLFGPQKPK VSMNQLSQQK PNQWLNQFVP PQGSPTMGSS VMGTQMNVIG QSAFGMQGNP FFNPQNFAQP PTTMTNSSSA SNDLKDLFG

          Target Information

          SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Coated vesicle-associated kinase of 104 kDa; SCY1-like 2; SCY1-like protein 2; SCY1-like, kinase-like 2; unnamed protein product

          View more View less

          Gene Aliases: AMC4; AMCNACC; CVAK104; KIAA1360; SCYL2

          View more View less

          UniProt ID: (Human) Q6P3W7

          View more View less

          Entrez Gene ID: (Human) 55681

          View more View less

          Function(s)
          protein kinase activity receptor binding protein binding ATP binding
          Process(es)
          brain development endosome to lysosome transport pyramidal neuron development receptor internalization clathrin-mediated endocytosis negative regulation of canonical Wnt signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline