Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • SEMA3C Antibodies

          Proteintech

          SEMA3C Polyclonal Antibody

          2 References
          View all (7) SEMA3C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SEMA3C Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SEMA3C Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SEMA3C Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SEMA3C Antibody (19242-1-AP) in WB

          Mouse heart tissue were subjected to SDS PAGE followed by western blot with 19242-1-AP (SEMA3C antibody) at dilution of 1:1000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SEMA3C Antibody in Western Blot (WB)
          SEMA3C Antibody in Western Blot (WB)
          SEMA3C Polyclonal Antibody

          Product Details

          19242-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SEMA3C Fusion Protein Ag5480 (34-380 aa encoded by BC030690)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          33 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: DELRETKTS EYFSLSHHPL DYRILLMDED QDRIYVGSKD HILSLNINNI SQEALSVFWP ASTIKVEECK MAGKDPTHGC GNFVRVIQTF NRTHLYVCGS GAFSPVCTYL NRGRRSEDQV FMIDSKCESG KGRCSFNPNV NTVSVMINEE LFSGMYIDFM GTDAAIFRSL TKRNAVRTDQ HNSKWLSEPM FVDAHVIPDG TDPNDAKVYF FFKEKLTDNN RSTKQIHSMI ARICPNDTGG LRSLVNKWTT FLKARLVCSV TDEDGPETHF DELEDVFLLE TDNPRTTLVY GIFTTSSSVF KGSAVCVYHL SDIQTVFNGP FAHKEGPNHQ LISYQGRIPY PRPGTCPG (34-380 aa encoded by BC030690 )

          Target Information

          The SEMA3C gene encodes the protein semaphorin 3C, a member of the semaphorin family known for its roles in axonal guidance, cellular signaling, and vascular development. Semaphorin 3C is involved in guiding neurons during development by serving as a repulsive cue, helping establish proper neural network formation through interaction with neuropilin and plexin receptors. It also plays crucial roles in angiogenesis and cardiac development by influencing endothelial cell behavior and migration. In the context of cancer, SEMA3C is implicated in tumor progression, where its dysregulation can promote tumor cell invasion and metastasis by modifying the extracellular matrix and affecting cellular adhesion. Research has demonstrated that SEMA3C can alter the tumor microenvironment, making it a potential target for cancer therapies aimed at inhibiting metastasis and controlling tumor growth. Understanding SEMA3C's function in various biological processes continues, highlighting its significance in developmental biology and disease modulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C; Sema E; semaphorin E; Semaphorin-3C; Semaphorin-E; unnamed protein product

          View more View less

          Gene Aliases: 1110036B02Rik; SEMA3C; SEMAE; SemE

          View more View less

          UniProt ID: (Human) Q99985, (Mouse) Q62181

          View more View less

          Entrez Gene ID: (Human) 10512, (Mouse) 20348

          View more View less

          Function(s)
          protein binding semaphorin receptor binding neuropilin binding chemorepellent activity membrane-bound signaling molecule intercellular signal molecule
          Process(es)
          neural crest cell migration somitogenesis blood vessel remodeling outflow tract septum morphogenesis outflow tract morphogenesis cardiac right ventricle morphogenesis pulmonary myocardium development immune response nervous system development axon guidance heart development response to xenobiotic stimulus post-embryonic development neural tube development cell differentiation positive regulation of cell migration negative chemotaxis limb bud formation dichotomous subdivision of terminal units involved in salivary gland branching semaphorin-plexin signaling pathway cardiac endothelial to mesenchymal transition positive regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline