Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SEMA4F Antibodies

          Abnova

          SEMA4F Polyclonal Antibody, MaxPab™

          View all (7) SEMA4F antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SEMA4F Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SEMA4F Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SEMA4F Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SEMA4F Antibody (H00010505-B01P) in WB

          Western Blot analysis of SEMA4F expression in transfected 293T cell line by SEMA4F MaxPab polyclonal antibody. Lane 1: SEMA4F transfected lysate(84.7 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SEMA4F Antibody in Western Blot (WB)
          SEMA4F Polyclonal Antibody, MaxPab™

          Product Details

          H00010505-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SEMA4F (ABM86603.1, 1 a.a. approximately 770 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPASAARPRP GPGQPTASPF PLLLLAVLSG PVSGRVPRSV PRTSLPISEA DSCLTRFAVP HTYNYSVLLV DPASHTLYVG ARDTIFALSL PFSGERPRRI DWMVPEAHRQ NCRKKGKKED ECHNFVQILA IANASHLLTC GTFAFDPKCG VIDVSRFQQV ERLESGRGKC PFEPAQRSAA VMAGGVLYAA TVKNYLGTEP IITRAVGRAE DWIRTDTLPS WLNAPAFVAA VALSPAEWGD EDGDDEIYFF FTETSRAFDS YERIKVPRVA RVCAGDLGGR KTLQQRWTTF LKADLLCPGP EHGRASSVLQ DVAVLRPELG AGTPIFYGIF SSQWEGATIS AVCAFRPQDI RTVLNGPFRE LKHDCNRGLP VVDNDVPQPR PGECITNNMK LRHFGSSLSL PDRVLTFIRD HPLMDRPVFP ADGHPLLVTT DTAYLRVVAH RVTSLSGKEY DVLYLGTEDG HLHRAVRIGA QLSVLEDLAL FPEPQPVENM KLYHSWLLVG SRTEVTQVNT TNCGRLQSCS ECILAQDPVC AWSFRLDECV AHAGEHRGLV QDIESADVSS LCPKEPGERP VVFEVPVATA AHVVLPCSPS SAWASCVWHQ PSGVTALTPR RDGLEVVVTP GAMGAYACEC QEGGAAHVVA AYSLVWGSQR DAPSRAHTVG AGLAGFFLGI LAASLTLILI GRRQQRRRQR ELLARDKVGL DLGAPPSGTT SYSQDPPSPS PEDERLPLAL AKRGSGFGGF SPPFLLDPCP SPAHIRLTGA PLATCDETSI

          Target Information

          SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F isoform SEMA4F_1; sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F isoform SEMA4F_2; sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F isoform SEMA4F_3; sema domain immunoglobulin domain transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 4F; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4F; Sema M; Sema W; semaphorin M; Semaphorin-4F; Semaphorin-M; Semaphorin-W; unnamed protein product

          View more View less

          Gene Aliases: M-SEMA; m-Sema-M; PRO2353; S4F; SEMA4F; SEMAM; SEMAW

          View more View less

          UniProt ID: (Human) O95754

          View more View less

          Entrez Gene ID: (Human) 10505

          View more View less

          Function(s)
          protein binding semaphorin receptor binding neuropilin binding chemorepellent activity
          Process(es)
          neural crest cell migration cell-cell signaling nervous system development axon guidance cell differentiation positive regulation of cell migration negative regulation of axon extension retinal ganglion cell axon guidance negative chemotaxis semaphorin-plexin signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline