Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total SEPT11.
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: septin 6; Septin-11
Gene Aliases: 6230410I01Rik; AW548875; D5Ertd606e; SEPT11; Sept6; SEPTIN11
UniProt ID: (Human) Q9NVA2, (Rat) B3GNI6, (Mouse) Q8C1B7
Entrez Gene ID: (Human) 55752, (Rat) 305227, (Mouse) 52398
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support