Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RTQRRKSTI TALLFGEDDL EALKAKNIKQ TELVADLREA ILRVARHFQC TDPKNCSVEL TPDYSMESHQ RDHENYVACS RSHRRRAKAL LDFERHDDDE LGFRKNDIIT IVSQKDEHCW VGELNGLRGW FPAKFVEVLD ERSKEYSIAG DDSVTEGVTD LVRGTLCPAL KALFEHGLKK PSLLGGACHP WLFIEEAAGR EVERDFASVY SRLVLCKTFR LDEDGKVLTP EELLYRAVQS VNVTHDAVHA QMDVKLRSLI CVGLNEQVLH LWLEVLCSSL PTVEKWYQPW SFLRSPGWVQ IKCELRVLCC FAFSLSQDWE LPAKREAQQP LKEGVRDMLV KHHLFSWDVD G (400-749 aa encoded by BC008078)
May play a cooperative role in NF2-mediated growth suppression of cells. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cx43-interacting protein of 85-kDa; dJ1042K10.2; DKFZp761D051; MAP; merlin binding protein; Merlin-associated protein; Rab-GTPase-activating protein-like protein; RabGAP-5; RabGAPLP; RUN and SH3 containing 3; RUN and TBC1 domain containing 3; RUN and TBC1 domain-containing protein 3; sgsm3; Small G protein signaling modulator 3; small G protein signaling modulator 3 protein; unnamed protein product
Gene Aliases: 1810012I01Rik; AI428557; BB175482; CIP85; MAP; R75178; RabGAP-5; RABGAP5; RABGAPLP; RUSC3; RUTBC3; SGSM3
UniProt ID: (Human) Q96HU1, (Mouse) Q8VCZ6
Entrez Gene ID: (Human) 27352, (Mouse) 105835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support