FIGURE: 1 / 2
Recommended positive controls: mouse brain, rat brain.
Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (87%), Rabbit (87%), Chicken (93%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Enables identical protein binding activity. Involved in negative regulation of blood-brain barrier permeability; negative regulation of gene expression; and negative regulation of protein phosphorylation. Located in perinuclear region of cytoplasm.