Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLAYCVQDAT VVDVEKRRNP SKHYVYIINV TWSDSTSQTI YRRYSKFFDL QMQLLDKFPI EGGQKDPKQR IIPFLPGKIL FRRSHIRDVA VKRLKPIDEY CRALVRLPPH ISQCDEVFRF FEARPEDVNP PKEDYGSSKR KSVWLSSWAE SPKKDVTGAD ATAEPMILEQ YVVVSNYKKQ ENSELSLQAG EVVDVIEKNE SGWWFVSTSE EQGWVPATYL EAQNGTRDDS DINTSKTGEE EKYVTVQPYT SQSKDEIGFE KGVTVEVIRK NLEGWWYIRY LGKEGWAPAS YLKKAKDDLP TRKKNLAGPV EIIGNIMEIS NLLNKKASGD KETPPAEGEG HEAPIAKKEI SLPILCNASN GSAVGVPDRT VSRLAQGSPA VARIAPQRAQ ISSPNLRTRP PPRRESSLGF QLPKPPEPPS VEVEYYTIAE FQSCISDGIS FRGGQKAEVI DKNSGGWWYV QIGEKEGWAP ASYIDKRKKP NLSRRTSTLT RPKVPPPAPP SKPKEAEEGP TGASESQDSP RKLKYEEPEY DIPAFGFDSE PELSEEPVED RASGERRPAQ PHRPSPASSL QRARFKVGES SEDVALEEET IYENEGFRPY AEDTLSARGS SGDSDSPGSS SLSLTRKNSP KSGSPKSSSL LKLKAEKNAQ AEMGKNHSSA SFSSSITINT TCCSSSSSSS SSLSKTSGDL KPRSASDAGI RGTPKVRAKK DADANAGLTS CPRAKPSVRP KPFLNRAESQ SQEKMDISTL RRQLRPTGQL RGGLKGSKSE DSELPPQTAS EAPSEGSRRS SSDLITLPAT TPPCPTKKEW EGPATSYMTC SAYQKVQDSE ISFPAGVEVQ VLEKQESGWW YVRFGELEGW APSHYLVLDE NEQPDPSGKE LDTVPAKGRQ NEGKSDSLEK IERRVQALNT VNQSKKATPP IPSKPPGGFG KTSGTPAVKM RNGVRQVAVR PQSVFVSPPP KDNNLSCALR RNESLTATDG LRGVRRNSSF STARSAAAEA KGRLAERAAS QGSDSPLLPA QRNSIPVSPV RPKPIEKSQF IHNNLKDVYV SIADYEGDEE TAGFQEGVSM EVLERNPNGW WYCQILDGVK PFKGWVPSNY LEKKN
Adapter protein involved in invadopodia and podosome formation, extracellular matrix degradation and invasiveness of some cancer cells. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. In association with ADAM12, mediates the neurotoxic effect of amyloid-beta peptide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein TKS5; adaptor protein TKS5; Five SH3 domain-containing protein; KIAA0418; SH3 and PX domain-containing protein 2A; SH3 multiple domains 1; SH3 multiple domains protein 1; Tyrosine kinase substrate with five SH3 domains; unnamed protein product
Gene Aliases: FISH; KIAA0418; SH3MD1; SH3PXD2A; TKS5
UniProt ID: (Human) Q5TCZ1
Entrez Gene ID: (Human) 9644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support