Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SHROOM3 Antibodies

          Abnova

          SHROOM3 Polyclonal Antibody, MaxPab™

          View all (3) SHROOM3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SHROOM3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SHROOM3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SHROOM3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SHROOM3 Antibody (H00057619-B01P) in WB

          Western Blot analysis of SHROOM3 expression in transfected 293T cell line by SHROOM3 MaxPab polyclonal antibody. Lane 1: SHROOM3 transfected lysate(219.5 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SHROOM3 Antibody in Western Blot (WB)

          Product Details

          H00057619-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SHROOM3 (AAI56133.1, 1 a.a. approximately 1995 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRTTEDFHKP SATLNSNTAT KGRYIYLEAF LEGGAPWGFT LKGGLEHGEP LIISKVEEGG KADTLSSKLQ AGDEVVHINE VTLSSSRKEA VSLVKGSYKT LRLVVRRDVC TDPGHADTGA SNFVSPEHLT SGPQHRKAAW SGGVKLRLKH RRSEPAGRPH SWHTTKSGEK QPDASMMQIS QGMIGPPWHQ SYHSSSSTSD LSNYDHAYLR RSPDQCSSQG SMESLEPSGA YPPCHLSPAK STGSIDQLSH FHNKRDSAYS SFSTSSSILE YPHPGISGRE RSGSMDNTSA RGGLLEGMRQ ADIRYVKTVY DTRRGVSAEY EVNSSALLLQ GREARASANG QGYDKWSNIP RGKGVPPPSW SQQCPSSLET ATDNLPPKVG APLPPARSDS YAAFRHRERP SSWSSLDQKR LCRPQANSLG SLKSPFIEEQ LHTVLEKSPE NSPPVKPKHN YTQKAQPGQP LLPTSIYPVP SLEPHFAQVP QPSVSSNGML YPALAKESGY IAPQGACNKM ATIDENGNQN GSGRPGFAFC QPLEHDLLSP VEKKPEATAK YVPSKVHFCS VPENEEDASL KRHLTPPQGN SPHSNERKST HSNKPSSHPH SLKCPQAQAW QAGEDKRSSR LSEPWEGDFQ EDHNANLWRR LEREGLGQSL SGNFGKTKSA FSSLQNIPES LRRHSSLELG RGTQEGYPGG RPTCAVNTKA EDPGRKAAPD LGSHLDRQVS YPRPEGRTGA SASFNSTDPS PEEPPAPSHP HTSSLGRRGP GPGSASALQG FQYGKPHCSV LEKVSKFEQR EQGSQRPSVG GSGFGHNYRP HRTVSTSSTS GNDFEETKAH IRFSESAEPL GNGEQHFKNG ELKLEEASRQ PCGQQLSGGA SDSGRGPQRP DARLLRSQST FQLSSEPERE PEWRDRPGSP ESPLLDAPFS RAYRNSIKDA QSRVLGATSF RRRDLELGAP VASRSWRPRP SSAHVGLRSP EASASASPHT PRERHSVTPA EGDLARPVPP AARRGARRRL TPEQKKRSYS EPEKMNEVGI VEEAEPAPLG PQRNGMRFPE SSVADRRRLF ERDGKACSTL SLSGPELKQF QQSALADYIQ RKTGKRPTSA AGCSLQEPGP LRERAQSAYL QPGPAALEGS GLASASSLSS LREPSLQPRR EATLLPATVA ETQQAPRDRS SSFAGGRRLG ERRRGDLLSG ANGGTRGTQR GDETPREPSS WGARAGKSMS AEDLLERSDV LAGPVHVRSR SSPATADKRQ DVLLGQDSGF GLVKDPCYLA GPGSRSLSCS ERGQEEMLPL FHHLTPRWGG SGCKAIGDSS VPSECPGTLD HQRQASRTPC PRPPLAGTQG LVTDTRAAPL TPIGTPLPSA IPSGYCSQDG QTGRQPLPPY TPAMMHRSNG HTLTQPPGPR GCEGDGPEHG VEEGTRKRVS LPQWPPPSRA KWAHAAREDS LPEESSAPDF ANLKHYQKQQ SLPSLCSTSD PDTPLGAPST PGRISLRISE SVLRDSPPPH EDYEDEVFVR DPHPKATSSP TFEPLPPPPP PPPSQETPVY SMDDFPPPPP HTVCEAQLDS EDPEGPRPSF NKLSKVTIAR ERHMPGAAHV VGSQTLASRL QTSIKGSEAE STPPSFMSVH AQLAGSLGGQ PAPIQTQSLS HDPVSGTQGL EKKVSPDPQK SSEDIRTEAL AKEIVHQDKS LADILDPDSR LKTTMDLMEG LFPRDVNLLK ENSVKRKAIQ RTVSSSGCEG KRNEDKEAVS MLVNCPAYYS VSAPKAELLN KIKEMPAEVN EEEEQADVNE KKAELIGSLT HKLETLQEAK GSLLTDIKLN NALGEEVEAL ISELCKPNEF DKYRMFIGDL DKVVNLLLSL SGRLARVENV LSGLGEDASN EERSSLYEKR KILAGQHEDA RELKENLDRR ERVVLGILAN YLSEEQLQDY QHFVKMKSTL LIEQRKLDDK IKLGQEQVKC LLESLPSDFI PKAGALALPP NLTSEPIPAG GCTFSGIFPT LTSPL

          Target Information

          SHROOM family members play diverse roles in the development of the nervous system and other tissues. SHROOM3 is an actin-binding protein is a regulator of the microtubule cytoskeleton during epithelial morphogenesis and is sufficient and necessary to induce a redistribution of the microtubule regulator gamma-tubulin. It is necessary for both apiscobasal cell elongation and apical constriction in the neural epithelium. SHROOM3 has also been shown to be essential for cell shape changes and morphogenesis in the developing vertebrate gut, with its transcription relying on the Pitx1 transcription factor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: F-actin-binding; F-actin-binding protein; hShrmL; Protein Shroom3; Shroom-related protein; unnamed protein product

          View more View less

          Gene Aliases: APXL3; KIAA1481; MSTP013; SHRM; SHRML; SHROOM3

          View more View less

          UniProt ID: (Human) Q8TF72

          View more View less

          Entrez Gene ID: (Human) 57619

          View more View less

          Function(s)
          actin binding actin filament binding
          Process(es)
          cell morphogenesis actin filament organization pattern specification process regulation of cell shape cellular pigment accumulation apical protein localization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline